Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 483112_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-760
          Stem-loop Accession Number: MI0005567
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 93846832 - 93846911 [+] on Build GRCh38
          Mature miRNA Sequence: CGGCUCUGGGUCUGUGGGGA
          Species: Human, Mouse, Rat, Bovine, Cricetulus griseus, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 483112_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000395, mir-760

          Mature miRNA Sequence:

          CGGCUCUGGGUCUGUGGGGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-760 MIMAT0004957
          Mouse mmu-miR-760-3p MIMAT0003898
          Rat rno-miR-760-3p MIMAT0005337
          Bovine bta-miR-760-3p MIMAT0022951
          Cricetulus griseus cgr-miR-760 MIMAT0041166
          Pan troglodytes ptr-miR-760 MIMAT0008319
          View More Rows
          Pongo pygmaeus ppy-miR-760 MIMAT0016104
          Rhesus monkey mml-miR-760 MIMAT0012788

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-760
          Stem-loop Accession # MI0005567
          Stem-loop Sequence
          GGCGCGUCGCCCCCCUCAGUCCACCAGAGCCCGGAUACCUCAGAAAUUCGGCUCUGGGUCUGUGGGGAGCGAAAUGCAAC
          Chromosome Location Chr. 1 - 93846832 - 93846911 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004957
          miRBase ID: hsa-miR-760
          Mature miRNA Sequence: CGGCUCUGGGUCUGUGGGGA
          Chromosome Location: Chr. 1 - 93846832 - 93846911 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-760
          Stem-loop Accession # MI0004605
          Stem-loop Sequence
          CGGGAGGAUGCCUCGGUGCGGGGCGCGUCGCCCCCCUCAGGCCACCAGAGCCCGGAUACCUCAGAAAUUCGGCUCUGGGUCUGUGGGGAGCGAAAUGCAACCCAAACCCCGUUUCCCCG
          Chromosome Location Chr. 3 - 122293585 - 122293703 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003898
          miRBase ID: mmu-miR-760-3p
          Mature miRNA Sequence: CGGCUCUGGGUCUGUGGGGA
          Chromosome Location: Chr. 3 - 122293585 - 122293703 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-760
          Stem-loop Accession # MI0006164
          Stem-loop Sequence
          GGCGCGUCGCCCCCCUCAGGCCACCAGAGCCCGGAUACCUCAGAAAUUCGGCUCUGGGUCUGUGGGGAGCGAAAUGCAAC
          Chromosome Location Chr. 2 - 225873354 - 225873433 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005337
          miRBase ID: rno-miR-760-3p
          Mature miRNA Sequence: CGGCUCUGGGUCUGUGGGGA
          Chromosome Location: Chr. 2 - 225873354 - 225873433 [-] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-760
          Stem-loop Accession # MI0009894
          Stem-loop Sequence
          GGAGGAUGCUGCAGCGUGGGGCGCGUCGCCCCCCUCAGUCCACCAGAGCCCGGAUACCUUAGAAAUUCGGCUCUGGGUCUGUGGGGAGCGAAAUGCAACCCAAACUCCAUUUUGCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022951
          miRBase ID: bta-miR-760-3p
          Mature miRNA Sequence: CGGCUCUGGGUCUGUGGGGA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-760
          Stem-loop Accession # MI0033096
          Stem-loop Sequence
          CGGGAGGAUGCCUCGGUGCGGGGCGCGUCGCCCCUCUCAGGCCACCAGAGCCCGGAGACCUCAGAAAUUCGGCUCUGGGUCUGUGGGGAGCGAAAUGCAACCCAAAGCCCGUUUCCCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041166
          miRBase ID: cgr-miR-760
          Mature miRNA Sequence: CGGCUCUGGGUCUGUGGGGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-760
          Stem-loop Accession # MI0008858
          Stem-loop Sequence
          GCGCGUCGCCCCCCUCAGUCCACCAGAGCCCGGAUACCUCAGAAAUUCGGCUCUGGGUCUGUGGGGAGCGAAAUGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008319
          miRBase ID: ptr-miR-760
          Mature miRNA Sequence: CGGCUCUGGGUCUGUGGGGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-760
          Stem-loop Accession # MI0015144
          Stem-loop Sequence
          GGCGCGUCGCCCCCCUCAGUCUACCAGAGCCCGGAUACCUCAGAAAUUCGGCUCUGGGUCUGUGGGGAGCGAAAUGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016104
          miRBase ID: ppy-miR-760
          Mature miRNA Sequence: CGGCUCUGGGUCUGUGGGGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-760
          Stem-loop Accession # MI0012545
          Stem-loop Sequence
          GGCGCGUCGCCCCCCUCAGUCCACCAGAGCCCGGAUACCUCAGAAAUUCGGCUCUGGGUCUGUGGGGAGCGAAAUGCAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012788
          miRBase ID: mml-miR-760
          Mature miRNA Sequence: CGGCUCUGGGUCUGUGGGGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03464716_pri
          TaqMan™ Pri-miRNA Assay : Hs03305191_pri
          TaqMan™ Pri-miRNA Assay : Mm03307938_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12767
          Ambion® Pre-miR™ miRNA Precursor : PM12767
          TaqMan™ MicroRNA Assay : 002328
          mirVana® miRNA inhibitor : MH12767
          mirVana® miRNA mimic : MC12767
          TaqMan™ Advanced miRNA Assay : rno481261_mir
          TaqMan™ Advanced miRNA Assay : mmu481261_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-vkppw:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0