Search
Search

| Assay Name: | hsa-miR-374a-5p |
|---|---|
| miRBase Accession Number: | MI0000782 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUAUAAUACAACCUGAUAAGUG |
| Species: | Human, Bovine, Dasypus novemcinctus, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Sheep, Tupaia chinensis |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000288, mir-374 |
Mature miRNA Sequence: | UUAUAAUACAACCUGAUAAGUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-374a-5p | MIMAT0000727 |
| Bovine | bta-miR-374a | MIMAT0004342 |
| Dasypus novemcinctus | dno-miR-374a-5p | MIMAT0047846 |
| Guinea pig | cpo-miR-374-5p | MIMAT0047205 |
| Horse | eca-miR-374a | MIMAT0013213 |
| Pan troglodytes | ptr-miR-374a | MIMAT0008123 |
| View More Rows | ||
| Pig | ssc-miR-374a-5p | MIMAT0013913 |
| Pongo pygmaeus | ppy-miR-374a | MIMAT0015862 |
| Rabbit | ocu-miR-374a-5p | MIMAT0048406 |
| Rhesus monkey | mml-miR-374a-5p | MIMAT0006300 |
| Sheep | oar-miR-374a | MIMAT0030062 |
| Tupaia chinensis | tch-miR-374a-5p | MIMAT0036589 |