Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 478238_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-374a-5p
          Stem-loop Accession Number: MI0000782
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 74287286 - 74287357 [-] on Build GRCh38
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Species: Human, Bovine, Dasypus novemcinctus, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Rabbit, Rhesus monkey, Sheep, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 478238_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000288, mir-374

          Mature miRNA Sequence:

          UUAUAAUACAACCUGAUAAGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-374a-5p MIMAT0000727
          Bovine bta-miR-374a MIMAT0004342
          Dasypus novemcinctus dno-miR-374a-5p MIMAT0047846
          Guinea pig cpo-miR-374-5p MIMAT0047205
          Horse eca-miR-374a MIMAT0013213
          Pan troglodytes ptr-miR-374a MIMAT0008123
          View More Rows
          Pig ssc-miR-374a-5p MIMAT0013913
          Pongo pygmaeus ppy-miR-374a MIMAT0015862
          Rabbit ocu-miR-374a-5p MIMAT0048406
          Rhesus monkey mml-miR-374a-5p MIMAT0006300
          Sheep oar-miR-374a MIMAT0030062
          Tupaia chinensis tch-miR-374a-5p MIMAT0036589

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-374a
          Stem-loop Accession # MI0000782
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUAUAGCACUUAUCAGAUUGUAUUGUAAUUGUCUGUGUA
          Chromosome Location Chr. X - 74287286 - 74287357 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000727
          miRBase ID: hsa-miR-374a-5p
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: Chr. X - 74287286 - 74287357 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-374a
          Stem-loop Accession # MI0005466
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGGUUGUAUUGUAAUUGUCUGUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004342
          miRBase ID: bta-miR-374a
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-374a
          Stem-loop Accession # MI0039088
          Stem-loop Sequence
          UUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGAUUGUAUUGUAAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047846
          miRBase ID: dno-miR-374a-5p
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-374
          Stem-loop Accession # MI0038751
          Stem-loop Sequence
          UUAUAAUACAACCUGAUAAGUGUUAUAACACUUACCAGAUUGUAUUGUAAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047205
          miRBase ID: cpo-miR-374-5p
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-374a
          Stem-loop Accession # MI0012959
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGGUUGUAUUGUAAUUGUCUGUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013213
          miRBase ID: eca-miR-374a
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-374a
          Stem-loop Accession # MI0008643
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUAUAGCACUUAUCAGAUUGUAUUGUAAUUGUCUGUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008123
          miRBase ID: ptr-miR-374a
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-374a
          Stem-loop Accession # MI0013130
          Stem-loop Sequence
          CAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGGUUGUAUUGUAAUUGUCUGUGUAUAUAUAUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013913
          miRBase ID: ssc-miR-374a-5p
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-374a
          Stem-loop Accession # MI0014919
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUGUAGCACUUAUCAGAUUGUAUUGUAAUUGUCUGUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015862
          miRBase ID: ppy-miR-374a
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-374a
          Stem-loop Accession # MI0039384
          Stem-loop Sequence
          UUAUAAUACAACCUGAUAAGUGUUACAGCACUUACCAGAUUGUAUUGUAAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048406
          miRBase ID: ocu-miR-374a-5p
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-374a
          Stem-loop Accession # MI0007718
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGAUUGUAUUGUAAUUGUCUGUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006300
          miRBase ID: mml-miR-374a-5p
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-374a
          Stem-loop Accession # MI0025283
          Stem-loop Sequence
          UACAUCGGCCAUUAUAAUACAACCUGAUAAGUGUUACAGCACUUAUCAGGUUGUAUUGUAAUUGUCUGUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030062
          miRBase ID: oar-miR-374a
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-374a
          Stem-loop Accession # MI0031255
          Stem-loop Sequence
          UUAUAAUACAACCUGAUAAGUGUUACAACACUUAUCAGAUUGUAUUGUCAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036589
          miRBase ID: tch-miR-374a-5p
          Mature miRNA Sequence: UUAUAAUACAACCUGAUAAGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304235_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10112
          Ambion® Pre-miR™ miRNA Precursor : PM10112
          TaqMan™ MicroRNA Assay : 000563
          mirVana® miRNA inhibitor : MH10112
          mirVana® miRNA mimic : MC10112
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Fast
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma Card
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma Card
          MIRNA : TaqMan® Advanced miRNA Human A 96-well Plates, Fast
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Standard
          FGMIR : TaqManTM Advanced miRNA Human A Card
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          FGMIR : TaqManTM Advanced miRNA Human A 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human Serum/Plasma 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human Serum/Plasma 96-well Plates, Fast

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline