Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10203
          Assay Name: hsa-miR-22-3p
          miRBase Accession Number: MI0000078
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 1713903 - 1713987 [-] on Build GRCh38
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Species: Human, Mouse, Rat, Artibeus jamaicensis, Ateles geoffroyi, Callithrix jacchus, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Guinea pig, Horse, Lagothrix lagotricha, Lemur catta, Macaca nemestrina, Microcebus murinus, Nomascus leucogenys, Ophiophagus hannah, Ornithorhynchus anatinus, Otolemur garnettii, Pan paniscus, Pan troglodytes, Papio hamadryas, Pig, Pongo pygmaeus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Saguinus labiatus, Saimiri boliviensis, Tupaia chinensis, Xenopus tropicalis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10203
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000053, mir-22

          Mature miRNA Sequence:

          AAGCUGCCAGUUGAAGAACUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-22-3p MIMAT0000077
          Mouse mmu-miR-22-3p MIMAT0000531
          Rat rno-miR-22-3p MIMAT0000791
          Artibeus jamaicensis aja-miR-22 MIMAT0031141
          Ateles geoffroyi age-miR-22 MIMAT0002327
          Callithrix jacchus cja-miR-22 MIMAT0049308
          View More Rows
          Chicken gga-miR-22-3p MIMAT0007288
          Chrysemys picta cpi-miR-22-3p MIMAT0037654
          Columba livia cli-miR-22-3p MIMAT0038439
          Cricetulus griseus cgr-miR-22-3p MIMAT0023857
          Dasypus novemcinctus dno-miR-22-3p MIMAT0047527
          Daubentonia madagascariensis dma-miR-22 MIMAT0049249
          Dog cfa-miR-22 MIMAT0006733
          Guinea pig cpo-miR-22-3p MIMAT0046875
          Horse eca-miR-22 MIMAT0013031
          Lagothrix lagotricha lla-miR-22 MIMAT0002334
          Lemur catta lca-miR-22 MIMAT0002329
          Macaca nemestrina mne-miR-22 MIMAT0002335
          Microcebus murinus mmr-miR-22 MIMAT0049184
          Nomascus leucogenys nle-miR-22 MIMAT0049355
          Ophiophagus hannah oha-miR-22a MIMAT0036845
          Ornithorhynchus anatinus oan-miR-22-3p MIMAT0006962
          Otolemur garnettii oga-miR-22 MIMAT0049697
          Pan paniscus ppa-miR-22 MIMAT0002328
          Pan troglodytes ptr-miR-22 MIMAT0002332
          Papio hamadryas pha-miR-22 MIMAT0049586
          Pig ssc-miR-22-3p MIMAT0015710
          Pongo pygmaeus ppy-miR-22 MIMAT0002331
          Pteropus alecto pal-miR-22-3p MIMAT0039861
          Python bivittatus pbv-miR-22-3p MIMAT0038852
          Rabbit ocu-miR-22-3p MIMAT0048127
          Rhesus monkey mml-miR-22 MIMAT0002330
          Saguinus labiatus sla-miR-22 MIMAT0002333
          Saimiri boliviensis sbo-miR-22 MIMAT0049496
          Tupaia chinensis tch-miR-22-3p MIMAT0036588
          Xenopus tropicalis xtr-miR-22-3p MIMAT0003672

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-22
          Stem-loop Accession # MI0000078
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location Chr. 17 - 1713903 - 1713987 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000077
          miRBase ID: hsa-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: Chr. 17 - 1713903 - 1713987 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-22
          Stem-loop Accession # MI0000570
          Stem-loop Sequence
          ACCUGGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCCCCUGGC
          Chromosome Location Chr. 11 - 75463716 - 75463810 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000531
          miRBase ID: mmu-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: Chr. 11 - 75463716 - 75463810 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-22
          Stem-loop Accession # MI0000851
          Stem-loop Sequence
          ACCUGGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCCACUGGC
          Chromosome Location Chr. 10 - 62299592 - 62299686 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000791
          miRBase ID: rno-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: Chr. 10 - 62299592 - 62299686 [+] on Build Rnor_6.0

          Artibeus jamaicensis

          Stem-loop ID aja-mir-22
          Stem-loop Accession # MI0024363
          Stem-loop Sequence
          ACCUGGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCCCCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031141
          miRBase ID: aja-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Ateles geoffroyi

          Stem-loop ID age-mir-22
          Stem-loop Accession # MI0002628
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002327
          miRBase ID: age-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-22
          Stem-loop Accession # MI0040044
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCCGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049308
          miRBase ID: cja-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-22
          Stem-loop Accession # MI0006982
          Stem-loop Sequence
          GCUCCUGGCUGCCCGGCAGCAGUUCUUCAGUGGCAAGCUUUAUGUCCUUCUCUAGUAGCUAAAGCUGCCAGUUGAAGAACUGUUGAAUGUAGCCACGUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007288
          miRBase ID: gga-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-22
          Stem-loop Accession # MI0029410
          Stem-loop Sequence
          CCGGCUAACUAGCACCAGUUCUUCAGUGGCAAGCUUUACGUCCCUGUCUAGCAGCUAAAGCUGCCAGUUGAAGAACUGUUGAAUGUAGCCACUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037654
          miRBase ID: cpi-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-22
          Stem-loop Accession # MI0029921
          Stem-loop Sequence
          CAGCACCAGUUCUUCAGUGGCAAGCUUUAUGUCCUUCUCUAGUAGCUAAAGCUGCCAGUUGAAGAACUGUUGAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038439
          miRBase ID: cli-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-22
          Stem-loop Accession # MI0020463
          Stem-loop Sequence
          GAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023857
          miRBase ID: cgr-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-22
          Stem-loop Accession # MI0038918
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047527
          miRBase ID: dno-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-22
          Stem-loop Accession # MI0039977
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049249
          miRBase ID: dma-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-22
          Stem-loop Accession # MI0008157
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006733
          miRBase ID: cfa-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-22
          Stem-loop Accession # MI0038574
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046875
          miRBase ID: cpo-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-22
          Stem-loop Accession # MI0012778
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013031
          miRBase ID: eca-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-22
          Stem-loop Accession # MI0002635
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002334
          miRBase ID: lla-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Lemur catta

          Stem-loop ID lca-mir-22
          Stem-loop Accession # MI0002630
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002329
          miRBase ID: lca-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-22
          Stem-loop Accession # MI0002636
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002335
          miRBase ID: mne-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-22
          Stem-loop Accession # MI0039903
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049184
          miRBase ID: mmr-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-22
          Stem-loop Accession # MI0040092
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049355
          miRBase ID: nle-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-22a
          Stem-loop Accession # MI0031425
          Stem-loop Sequence
          CUACCCGCAGCAGUUCUUCAGUGGCACGCUUUACGUCCCCUCCAGCAACUAAAGCUGCCAGUUGAAGAACUGUUGGCCGUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036845
          miRBase ID: oha-miR-22a
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-22
          Stem-loop Accession # MI0006761
          Stem-loop Sequence
          CCCGUGUGCCGGCCCGGGCCUGGCUGAGUCACAGCAGUUCUUCAGUGGCAAGCUUUACGUCCUGUCCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGAACUCAGCCCCUGUCUGCCACGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006962
          miRBase ID: oan-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-22
          Stem-loop Accession # MI0040481
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049697
          miRBase ID: oga-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-22
          Stem-loop Accession # MI0002629
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002328
          miRBase ID: ppa-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-22
          Stem-loop Accession # MI0002633
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002332
          miRBase ID: ptr-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-22
          Stem-loop Accession # MI0040364
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049586
          miRBase ID: pha-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-22
          Stem-loop Accession # MI0014770
          Stem-loop Sequence
          GAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015710
          miRBase ID: ssc-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-22
          Stem-loop Accession # MI0002632
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002331
          miRBase ID: ppy-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-22
          Stem-loop Accession # MI0032368
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039861
          miRBase ID: pal-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-22
          Stem-loop Accession # MI0030169
          Stem-loop Sequence
          UGGCUACCCAGCAGCAGUUCUUCAGUGGCACGCUUUACGUCCCCUCCAGCAGCUAAAGCUGCCAGUUGAAGAACUGUUGACCGUAGCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038852
          miRBase ID: pbv-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-22
          Stem-loop Accession # MI0039234
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048127
          miRBase ID: ocu-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-22
          Stem-loop Accession # MI0002631
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002330
          miRBase ID: mml-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-22
          Stem-loop Accession # MI0002634
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCCGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002333
          miRBase ID: sla-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Saimiri boliviensis

          Stem-loop ID sbo-mir-22
          Stem-loop Accession # MI0040256
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCCGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049496
          miRBase ID: sbo-miR-22
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-22
          Stem-loop Accession # MI0031254
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036588
          miRBase ID: tch-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-22
          Stem-loop Accession # MI0004912
          Stem-loop Sequence
          CUGGCCCGCUAGAAGCAGUUCUUCAGUGGCAAGCUUUAUGUUGUUCCUCUGUGCUAAAGCUGCCAGUUGAAGAACUGUUGAAAGUGGCUACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003672
          miRBase ID: xtr-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302632_pri
          TaqMan™ Pri-miRNA Assay : Rn03465003_pri
          TaqMan™ Pri-miRNA Assay : Mm03306829_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10203
          TaqMan™ MicroRNA Assay : 000398
          mirVana® miRNA inhibitor : MH10203
          mirVana® miRNA mimic : MC10203
          TaqMan™ Advanced miRNA Assay : mmu481004_mir
          TaqMan™ Advanced miRNA Assay : rno481004_mir
          TaqMan™ Advanced miRNA Assay : 477985_mir


          miRBase Alias:

          Human: hsa-miR-22 (v17)
          Mouse: mmu-miR-22 (v17)
          Rat: rno-miR-22 (v18)
          Gallus gallus: gga-miR-22 (v18)
          Ochrobactrum anthropi: oan-miR-22 (v18)
          Xenopus tropicalis: xtr-miR-22 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-86tfw:80/100.66.78.224:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0