Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn03465003_pri
          Assay Name:
          rno-mir-22
          Stem-loop ID:
          rno-mir-22 rno-mir-6326
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.10: 62299592 - 62299686 [+] on Build Rnor_6.0
          Stem-loop Location:
          Chr.10: 62299592 - 62299686 [+] on Build Rnor_6.0 Chr.10: 62299339 - 62299449 [+] on Build Rnor_6.0
          Mature miRNA ID:
          rno-miR-22-3p rno-miR-22-5p rno-miR-6326
          Species:
          Rat
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Rn03465003_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          62

          Chromosome Location:

          Chr. 10 - 62299841 - 62299841 [+] on 62299841 Build Rnor_6.0

          Species:

          Rat

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-22-3p MIMAT0000791
          Rat rno-miR-22-5p MIMAT0003152
          Rat rno-miR-6326 MIMAT0025065

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-22
          Stem-loop Accession # MI0000851
          Stem-loop Sequence
          ACCUGGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCCACUGGC
          Chromosome Location Chr. 10 - 62299592 - 62299686 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000791
          miRBase ID: rno-miR-22-3p
          Mature miRNA Sequence: AAGCUGCCAGUUGAAGAACUGU
          Chromosome Location: Chr. 10 - 62299592 - 62299686 [+] on Build Rnor_6.0

          miRBase Accession #: MIMAT0003152
          miRBase ID: rno-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: Chr. 10 - 62299592 - 62299686 [+] on Build Rnor_6.0

          Gene Family ID MIPF0000053, mir-22
          Distance to Amplicon 124 bases
          Stem-loop ID rno-mir-6326
          Stem-loop Accession # MI0021850
          Stem-loop Sequence
          CUGCCCUUUGAGUGCUGAAGGUGCUGGAGUCCUUAGCAGGUUGCCAAGAGUUGCUAGUGCCUCCUGGUCUGUAAAGGGACACAGCACAAGUGCAUUUAGGAGCCUCUUGCU
          Chromosome Location Chr. 10 - 62299339 - 62299449 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0025065
          miRBase ID: rno-miR-6326
          Mature miRNA Sequence: CUGGUCUGUAAAGGGACACAGCA
          Chromosome Location: Chr. 10 - 62299339 - 62299449 [+] on Build Rnor_6.0

          Gene Family ID
          Distance to Amplicon 361 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM24771
          Ambion® Pre-miR™ miRNA Precursor : PM24771
          TaqMan™ MicroRNA Assay : 476908_mat
          mirVana® miRNA inhibitor : MH24771
          mirVana® miRNA mimic : MC24771
          TaqMan™ Advanced miRNA Assay : rno481234_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM10203
          Ambion® Pre-miR™ miRNA Precursor : PM10203
          TaqMan™ MicroRNA Assay : 000398
          mirVana® miRNA inhibitor : MH10203
          mirVana® miRNA mimic : MC10203
          TaqMan™ Advanced miRNA Assay : mmu481004_mir
          TaqMan™ Advanced miRNA Assay : rno481004_mir
          TaqMan™ Advanced miRNA Assay : 477985_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM11752
          Ambion® Pre-miR™ miRNA Precursor : PM11752
          TaqMan™ MicroRNA Assay : 002301
          mirVana® miRNA inhibitor : MH11752
          mirVana® miRNA mimic : MC11752
          TaqMan™ Advanced miRNA Assay : rno477987_mir
          TaqMan™ Advanced miRNA Assay : mmu482711_mir
          TaqMan™ Advanced miRNA Assay : 477987_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-86tfw:80/100.66.78.224:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0