Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM11752
          Assay Name: hsa-miR-22-5p
          miRBase Accession Number: MI0000078
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 1713903 - 1713987 [-] on Build GRCh38
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Species: Human, Mouse, Rat, Bovine, Chicken, Chrysemys picta, Columba livia, Dasypus novemcinctus, Guinea pig, Ornithorhynchus anatinus, Pig, Pteropus alecto, Rabbit
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM11752
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000053, mir-22

          Mature miRNA Sequence:

          AGUUCUUCAGUGGCAAGCUUUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-22-5p MIMAT0004495
          Mouse mmu-miR-22-5p MIMAT0004629
          Rat rno-miR-22-5p MIMAT0003152
          Bovine bta-miR-22-5p MIMAT0003826
          Chicken gga-miR-22-5p MIMAT0007287
          Chrysemys picta cpi-miR-22-5p MIMAT0037653
          View More Rows
          Columba livia cli-miR-22-5p MIMAT0038438
          Dasypus novemcinctus dno-miR-22-5p MIMAT0047526
          Guinea pig cpo-miR-22-5p MIMAT0046874
          Ornithorhynchus anatinus oan-miR-22-5p MIMAT0006961
          Pig ssc-miR-22-5p MIMAT0015709
          Pteropus alecto pal-miR-22-5p MIMAT0039860
          Rabbit ocu-miR-22-5p MIMAT0048126

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-22
          Stem-loop Accession # MI0000078
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location Chr. 17 - 1713903 - 1713987 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004495
          miRBase ID: hsa-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: Chr. 17 - 1713903 - 1713987 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-22
          Stem-loop Accession # MI0000570
          Stem-loop Sequence
          ACCUGGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCCCCUGGC
          Chromosome Location Chr. 11 - 75463716 - 75463810 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004629
          miRBase ID: mmu-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: Chr. 11 - 75463716 - 75463810 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-22
          Stem-loop Accession # MI0000851
          Stem-loop Sequence
          ACCUGGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCCACUGGC
          Chromosome Location Chr. 10 - 62299592 - 62299686 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003152
          miRBase ID: rno-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: Chr. 10 - 62299592 - 62299686 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-22
          Stem-loop Accession # MI0005041
          Stem-loop Sequence
          GGCUGAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003826
          miRBase ID: bta-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-22
          Stem-loop Accession # MI0006982
          Stem-loop Sequence
          GCUCCUGGCUGCCCGGCAGCAGUUCUUCAGUGGCAAGCUUUAUGUCCUUCUCUAGUAGCUAAAGCUGCCAGUUGAAGAACUGUUGAAUGUAGCCACGUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007287
          miRBase ID: gga-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-22
          Stem-loop Accession # MI0029410
          Stem-loop Sequence
          CCGGCUAACUAGCACCAGUUCUUCAGUGGCAAGCUUUACGUCCCUGUCUAGCAGCUAAAGCUGCCAGUUGAAGAACUGUUGAAUGUAGCCACUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037653
          miRBase ID: cpi-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-22
          Stem-loop Accession # MI0029921
          Stem-loop Sequence
          CAGCACCAGUUCUUCAGUGGCAAGCUUUAUGUCCUUCUCUAGUAGCUAAAGCUGCCAGUUGAAGAACUGUUGAAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038438
          miRBase ID: cli-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-22
          Stem-loop Accession # MI0038918
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047526
          miRBase ID: dno-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-22
          Stem-loop Accession # MI0038574
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046874
          miRBase ID: cpo-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-22
          Stem-loop Accession # MI0006761
          Stem-loop Sequence
          CCCGUGUGCCGGCCCGGGCCUGGCUGAGUCACAGCAGUUCUUCAGUGGCAAGCUUUACGUCCUGUCCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGAACUCAGCCCCUGUCUGCCACGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006961
          miRBase ID: oan-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-22
          Stem-loop Accession # MI0014770
          Stem-loop Sequence
          GAGCCGCAGUAGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGUUGCCCUCUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015709
          miRBase ID: ssc-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-22
          Stem-loop Accession # MI0032368
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039860
          miRBase ID: pal-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-22
          Stem-loop Accession # MI0039234
          Stem-loop Sequence
          AGUUCUUCAGUGGCAAGCUUUAUGUCCUGACCCAGCUAAAGCUGCCAGUUGAAGAACUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048126
          miRBase ID: ocu-miR-22-5p
          Mature miRNA Sequence: AGUUCUUCAGUGGCAAGCUUUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302632_pri
          TaqMan™ Pri-miRNA Assay : Rn03465003_pri
          TaqMan™ Pri-miRNA Assay : Mm03306829_pri
          Ambion® Pre-miR™ miRNA Precursor : PM11752
          TaqMan™ MicroRNA Assay : 002301
          mirVana® miRNA inhibitor : MH11752
          mirVana® miRNA mimic : MC11752
          TaqMan™ Advanced miRNA Assay : rno477987_mir
          TaqMan™ Advanced miRNA Assay : mmu482711_mir
          TaqMan™ Advanced miRNA Assay : 477987_mir


          miRBase Alias:

          Human: hsa-miR-22* (v17)
          Mouse: mmu-miR-22* (v17)
          Rat: rno-miR-22* (v18)
          Gallus gallus: gga-miR-22* (v18)
          Ochrobactrum anthropi: oan-miR-22* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5d6d8bc979-86tfw:80/100.66.78.224:80.
          git-commit: e71199a02a6c1449b2ac9ded8316d5734e8aa2de
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.62.0-2026.11.45-1.0