Search
Search

| Assay Name: | hsa-miR-212-3p |
|---|---|
| miRBase Accession Number: | MI0000288 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAACAGUCUCCAGUCACGGCC |
| Species: | Human, Horse, Monodelphis domestica, Nomascus leucogenys |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000065, mir-132 |
Mature miRNA Sequence: | UAACAGUCUCCAGUCACGGCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-212-3p | MIMAT0000269 |
| Horse | eca-miR-212 | MIMAT0013030 |
| Monodelphis domestica | mdo-miR-212-3p | MIMAT0004148 |
| Nomascus leucogenys | nle-miR-212 | MIMAT0049353 |