Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03302957_pri
          Assay Name:
          hsa-mir-212
          Stem-loop ID:
          hsa-mir-212
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.17: 2050271 - 2050380 [-] on Build GRCh38
          Stem-loop Location:
          Chr.17: 2050271 - 2050380 [-] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-212-3p hsa-miR-212-5p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs03302957_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          85

          Chromosome Location:

          Chr. 17 - 2050767 - 2050767 [+] on 2050767 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-212-3p MIMAT0000269
          Human hsa-miR-212-5p MIMAT0022695

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-212
          Stem-loop Accession # MI0000288
          Stem-loop Sequence
          CGGGGCACCCCGCCCGGACAGCGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCACCGACGCCUGGCCCCGCC
          Chromosome Location Chr. 17 - 2050271 - 2050380 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000269
          miRBase ID: hsa-miR-212-3p
          Mature miRNA Sequence: UAACAGUCUCCAGUCACGGCC
          Chromosome Location: Chr. 17 - 2050271 - 2050380 [-] on Build GRCh38

          miRBase Accession #: MIMAT0022695
          miRBase ID: hsa-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: Chr. 17 - 2050271 - 2050380 [-] on Build GRCh38

          Gene Family ID MIPF0000065, mir-132
          Distance to Amplicon 329 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM10340
          Ambion® Pre-miR™ miRNA Precursor : PM10340
          TaqMan™ MicroRNA Assay : 000515
          mirVana® miRNA inhibitor : MH10340
          mirVana® miRNA mimic : MC10340
          TaqMan™ Advanced miRNA Assay : 478318_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM13202
          Ambion® Pre-miR™ miRNA Precursor : PM13202
          TaqMan™ MicroRNA Assay : 461768_mat
          mirVana® miRNA inhibitor : MH13202
          mirVana® miRNA mimic : MC13202
          TaqMan™ Advanced miRNA Assay : mmu482704_mir
          TaqMan™ Advanced miRNA Assay : 478767_mir

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline