Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13202
          Assay Name: hsa-miR-212-5p
          miRBase Accession Number: MI0000288
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 2050271 - 2050380 [-] on Build GRCh38
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Species: Human, Mouse, Bovine, Chicken, Cricetulus griseus, Dog, Guinea pig, Ictalurus punctatus, Monodelphis domestica, Neolamprologus brichardi, Pig, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Zebrafish
          Product Type: mirVana® miRNA mimic
          Assay ID MC13202

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000065, mir-132

          Mature miRNA Sequence:

          ACCUUGGCUCUAGACUGCUUACU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-212-5p MIMAT0022695
          Mouse mmu-miR-212-5p MIMAT0017053
          Bovine bta-miR-212 MIMAT0009264
          Chicken gga-miR-212-5p MIMAT0049954
          Cricetulus griseus cgr-miR-212 MIMAT0023850
          Dog cfa-miR-212 MIMAT0006731
          View More Rows
          Guinea pig cpo-miR-212-5p MIMAT0047113
          Ictalurus punctatus ipu-miR-212 MIMAT0029465
          Monodelphis domestica mdo-miR-212-5p MIMAT0026692
          Neolamprologus brichardi nbr-miR-212b MIMAT0042620
          Pig ssc-miR-212 MIMAT0025370
          Pteropus alecto pal-miR-212-5p MIMAT0039979
          Python bivittatus pbv-miR-212-5p MIMAT0039016
          Rabbit ocu-miR-212-5p MIMAT0048335
          Rhesus monkey mml-miR-212-5p MIMAT0026847
          Salmo salar ssa-miR-212b-5p MIMAT0032512
          Zebrafish dre-miR-212-5p MIMAT0048650

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-212
          Stem-loop Accession # MI0000288
          Stem-loop Sequence
          CGGGGCACCCCGCCCGGACAGCGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCACCGACGCCUGGCCCCGCC
          Chromosome Location Chr. 17 - 2050271 - 2050380 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0022695
          miRBase ID: hsa-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: Chr. 17 - 2050271 - 2050380 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-212
          Stem-loop Accession # MI0000696
          Stem-loop Sequence
          GGGCAGCGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCUUCAGUAACAGUCUCCAGUCACGGCCACCGACGCCUGGCCC
          Chromosome Location Chr. 11 - 75173388 - 75173478 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017053
          miRBase ID: mmu-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: Chr. 11 - 75173388 - 75173478 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-212
          Stem-loop Accession # MI0009776
          Stem-loop Sequence
          CGGAGCCACCCUGCCCGGGCAGCGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCACCGACGCCUGGCCCCGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009264
          miRBase ID: bta-miR-212
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-mir-212
          Stem-loop Accession # MI0040694
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCUGCGUAGGGACAGGACAGUAACAGUCUACAGUCAUGGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049954
          miRBase ID: gga-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-212
          Stem-loop Accession # MI0020459
          Stem-loop Sequence
          CGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023850
          miRBase ID: cgr-miR-212
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-212
          Stem-loop Accession # MI0008155
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006731
          miRBase ID: cfa-miR-212
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-212
          Stem-loop Accession # MI0038704
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047113
          miRBase ID: cpo-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-212-1
          Stem-loop Accession # MI0024570
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCUAAAUGUGUCUGAAAGUACAGUAACAGUCUACAGUCAUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029465
          miRBase ID: ipu-miR-212
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Stem-loop ID ipu-mir-212-2
          Stem-loop Accession # MI0024571
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCUUAUGGAGUCCGACAGUACAGUAACAGUCUACAGUCAUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029465
          miRBase ID: ipu-miR-212
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-212
          Stem-loop Accession # MI0005337
          Stem-loop Sequence
          GGGCAGCGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCACCCUCAGUAACAGUCUCCAGUCACGGCCACCGACGCCUGGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026692
          miRBase ID: mdo-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-212b
          Stem-loop Accession # MI0034499
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCUAAAACUCCUCCAAAGUACAGUAACAGUCUACAGUCAUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042620
          miRBase ID: nbr-miR-212b
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-212
          Stem-loop Accession # MI0022140
          Stem-loop Sequence
          GCGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCACCGACGCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025370
          miRBase ID: ssc-miR-212
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-212
          Stem-loop Accession # MI0032431
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039979
          miRBase ID: pal-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-212
          Stem-loop Accession # MI0030286
          Stem-loop Sequence
          CGGGACCUUGGCUCUAGACUGCUUACUGCUACAGGCAAUGAGAGACACAGUAACAGUCUACAGUCAUGGCUACUGAAGUCUGGCAGUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039016
          miRBase ID: pbv-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-212
          Stem-loop Accession # MI0039347
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCACGGCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048335
          miRBase ID: ocu-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-212
          Stem-loop Accession # MI0007671
          Stem-loop Sequence
          CGGGGCACCCCGCCCGGACAGCGCGCCGGCACCUUGGCUCUAGACUGCUUACUGCCCGGGCCGCCCUCAGUAACAGUCUCCAGUCAGGGCCACCGACGCCUGGCCCCGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026847
          miRBase ID: mml-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-212b-1
          Stem-loop Accession # MI0026609
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCUUAAAACUCAGUGAAACUACAGUAACAGUCUACAGUCAUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032512
          miRBase ID: ssa-miR-212b-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Stem-loop ID ssa-mir-212b-2
          Stem-loop Accession # MI0026610
          Stem-loop Sequence
          ACCUUGGCUCUAGACUGCUUACUGCUUAAAACUCAUUGUAACAGUCUACAGUCAUGGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032512
          miRBase ID: ssa-miR-212b-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-212-2
          Stem-loop Accession # MI0039509
          Stem-loop Sequence
          UCAGAACUUCAUGACCUUGGCUCUAGACUGCUUACUGCUUAUGGAGUCCGACAGUACAGUAACAGUCUACAGUCAUGGCUACUGAAGUCUGGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048650
          miRBase ID: dre-miR-212-5p
          Mature miRNA Sequence: ACCUUGGCUCUAGACUGCUUACU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302957_pri
          TaqMan™ Pri-miRNA Assay : Mm03307114_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13202
          Ambion® Pre-miR™ miRNA Precursor : PM13202
          TaqMan™ MicroRNA Assay : 461768_mat
          mirVana® miRNA inhibitor : MH13202
          TaqMan™ Advanced miRNA Assay : mmu482704_mir
          TaqMan™ Advanced miRNA Assay : 478767_mir

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline