Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10568
          Assay Name:
          miRBase Accession Number: MI0007155
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Species: Chicken, Ciona intestinalis, Heliconius melpomene, Monodelphis domestica, Oikopleura dioica, Xenopus tropicalis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10568
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000022, mir-7

          Mature miRNA Sequence:

          UGGAAGACUAGUGAUUUUGUUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Chicken gga-miR-7 MIMAT0001157
          Ciona intestinalis cin-miR-7-5p MIMAT0006091
          Heliconius melpomene hme-miR-7 MIMAT0024984
          Monodelphis domestica mdo-miR-7 MIMAT0004086
          Oikopleura dioica odi-miR-7 MIMAT0006078
          Xenopus tropicalis xtr-miR-7 MIMAT0003552

          Stem-loop Details



          Chicken

          Stem-loop ID gga-mir-7-1
          Stem-loop Accession # MI0001272
          Stem-loop Sequence
          UGGAUGUUGGUCUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAAUUAAAUUGACAACAAAUCACAGUCUGCCAUAUGGCACAGAUCAUGCCUCUACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001157
          miRBase ID: gga-miR-7
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA

          Stem-loop ID gga-mir-7-2
          Stem-loop Accession # MI0001226
          Stem-loop Sequence
          GGACGGCCGCGGUGCCCUCUGGAAGACUAGUGAUUUUGUUGUUGUAUGGCUCAUCCCACCACAACAAGUCACAGUCUGCCUUAGGGCGCACGGCCCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001157
          miRBase ID: gga-miR-7
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA

          Stem-loop ID gga-mir-7-3
          Stem-loop Accession # MI0001269
          Stem-loop Sequence
          CUGUGGUCUGGCUCUGUGUGGAAGACUAGUGAUUUUGUUGUUAUGAUUUAUAAAGGUGACAACAAAUCAUAGCCUGCCAUACAGCACAGAUCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001157
          miRBase ID: gga-miR-7
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA

          Ciona intestinalis

          Stem-loop ID cin-mir-7
          Stem-loop Accession # MI0007155
          Stem-loop Sequence
          AUAUCCGCACCCGGUUGGAAGACUAGUGAUUUUGUUGUUAUAGCAACAACAAUCUAUGGUCUCCUGCACGGAAGUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006091
          miRBase ID: cin-miR-7-5p
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA

          Heliconius melpomene

          Stem-loop ID hme-mir-7
          Stem-loop Accession # MI0021781
          Stem-loop Sequence
          UCCACGUGACUAACAGCGAUCUGCCUCGCGUUGUAUGGAAGACUAGUGAUUUUGUUGUUUGAUAAUAUAUAUAACAAUAAAUCACUAAUCUCCCUACAAAGCGCUGGCACCCUCUUCGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024984
          miRBase ID: hme-miR-7
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-7
          Stem-loop Accession # MI0005270
          Stem-loop Sequence
          UUGGCCUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAACGACAACAAAUCACAGUCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004086
          miRBase ID: mdo-miR-7
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA

          Oikopleura dioica

          Stem-loop ID odi-mir-7-1
          Stem-loop Accession # MI0007140
          Stem-loop Sequence
          AUCUGGUUGACGGUUUGGAAGACUAGUGAUUUUGUUGACUUGGACAUAAUUGUCGUUUCUUGUCAACUCACACUAGUUUCCCGUACUGCAUCGGAUUCAAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006078
          miRBase ID: odi-miR-7
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA

          Stem-loop ID odi-mir-7-2
          Stem-loop Accession # MI0007141
          Stem-loop Sequence
          AUCUGGUUGACGGUUUGGAAGACUAGUGAUUUUGUUGACUUGGACAUAAUUGUCGUUUCUUGUCAACUCACACUAGUUUCCCGUACUGCAUCGGAUUCAAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006078
          miRBase ID: odi-miR-7
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-7-1
          Stem-loop Accession # MI0004790
          Stem-loop Sequence
          UGGAUGUUGGUUUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACAACAUUGACAACAAAUCGCAGUCUGCCAUAUGGCACAGACCAUGCCUCUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003552
          miRBase ID: xtr-miR-7
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA

          Stem-loop ID xtr-mir-7-2
          Stem-loop Accession # MI0004792
          Stem-loop Sequence
          GAGGUGCAGGCUGACUCUUUGUGGAAGACUAGUGAUUUUGUUGUUGUAAGCCUUAUUGCAUGACAACAAGUCACAGUCUGCCUCACAGUGCCCAGCAAUAUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003552
          miRBase ID: xtr-miR-7
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA

          Stem-loop ID xtr-mir-7-3
          Stem-loop Accession # MI0004791
          Stem-loop Sequence
          CUGGGUCGGUUUGGAAGACUAGUGAUUUUGUUGUUUUUAUAAAAGUUUGUCACAAACAGCAAAUCGUAGUCUCCACUCUGCCCCAGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003552
          miRBase ID: xtr-miR-7
          Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Pre-miR™ miRNA Precursor : PM10568
          TaqMan™ MicroRNA Assay : 000386
          mirVana® miRNA inhibitor : MH10568
          mirVana® miRNA mimic : MC10568


          miRBase Alias:

          Human: hsa-miR-7 (v9.2)
          Mouse: mmu-miR-7 (v9.2)
          Chryseobacterium indologenes: cin-miR-7 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline