Search
Search

| Assay Name: | cin-miR-7-5p |
|---|---|
| miRBase Accession Number: | MI0007155 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAAGACUAGUGAUUUUGUUG |
| Species: | Chicken, Ciona intestinalis, Heliconius melpomene, Monodelphis domestica, Oikopleura dioica, Xenopus tropicalis |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000022, mir-7 |
Mature miRNA Sequence: | UGGAAGACUAGUGAUUUUGUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-7 | MIMAT0001157 |
| Ciona intestinalis | cin-miR-7-5p | MIMAT0006091 |
| Heliconius melpomene | hme-miR-7 | MIMAT0024984 |
| Monodelphis domestica | mdo-miR-7 | MIMAT0004086 |
| Oikopleura dioica | odi-miR-7 | MIMAT0006078 |
| Xenopus tropicalis | xtr-miR-7 | MIMAT0003552 |