Search
Search

| Assay Name: | hsa-miR-154-3p |
|---|---|
| miRBase Accession Number: | MI0000480 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAUCAUACACGGUUGACCUAUU |
| Species: | Human, Mouse, Rat, Cricetulus griseus, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUCAUACACGGUUGACCUAUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-154-3p | MIMAT0000453 |
| Mouse | mmu-miR-154-3p | MIMAT0004537 |
| Rat | rno-miR-154-3p | MIMAT0017136 |
| Cricetulus griseus | cgr-miR-154-3p | MIMAT0023787 |
| Rhesus monkey | mml-miR-154-3p | MIMAT0026830 |