Search
Search

Amplicon Length: | 102 |
Chromosome Location: | Chr. 6 - 133889030 - 133889030 [+] on 133889030 Build Rnor_6.0 |
Species: | Rat |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-154-3p | MIMAT0017136 |
| Rat | rno-miR-154-5p | MIMAT0000856 |
| Rat | rno-miR-496-3p | MIMAT0012860 |
| Rat | rno-miR-496-5p | MIMAT0017372 |