Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10086
          Assay Name: hsa-miR-154-5p
          miRBase Accession Number: MI0000480
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101059755 - 101059838 [+] on Build GRCh38
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Species: Human, Mouse, Rat, Cricetulus griseus, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Macaca nemestrina, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Rabbit, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC10086

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000018, mir-154

          Mature miRNA Sequence:

          UAGGUUAUCCGUGUUGCCUUCG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-154-5p MIMAT0000452
          Mouse mmu-miR-154-5p MIMAT0000164
          Rat rno-miR-154-5p MIMAT0000856
          Cricetulus griseus cgr-miR-154-5p MIMAT0023786
          Dasypus novemcinctus dno-miR-154a-5p MIMAT0047683
          Gorilla gorilla ggo-miR-154 MIMAT0002297
          View More Rows
          Guinea pig cpo-miR-154-5p MIMAT0047036
          Horse eca-miR-154a MIMAT0013129
          Macaca nemestrina mne-miR-154 MIMAT0002299
          Pan paniscus ppa-miR-154 MIMAT0002300
          Pan troglodytes ptr-miR-154 MIMAT0002296
          Pongo pygmaeus ppy-miR-154 MIMAT0002298
          Rabbit ocu-miR-154-5p MIMAT0048271
          Rhesus monkey mml-miR-154-5p MIMAT0006215

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-154
          Stem-loop Accession # MI0000480
          Stem-loop Sequence
          GUGGUACUUGAAGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUUUGUGACGAAUCAUACACGGUUGACCUAUUUUUCAGUACCAA
          Chromosome Location Chr. 14 - 101059755 - 101059838 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000452
          miRBase ID: hsa-miR-154-5p
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: Chr. 14 - 101059755 - 101059838 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-154
          Stem-loop Accession # MI0000176
          Stem-loop Sequence
          GAAGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUUCGUGACGAAUCAUACACGGUUGACCUAUUUUU
          Chromosome Location Chr. 12 - 109738433 - 109738498 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000164
          miRBase ID: mmu-miR-154-5p
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: Chr. 12 - 109738433 - 109738498 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-154
          Stem-loop Accession # MI0000923
          Stem-loop Sequence
          GCGGUGCUUGAAGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUUCGUGACGAAUCAUACACGGUUGACCUAUUUUUCAGUACCAA
          Chromosome Location Chr. 6 - 133888586 - 133888669 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000856
          miRBase ID: rno-miR-154-5p
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: Chr. 6 - 133888586 - 133888669 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-154
          Stem-loop Accession # MI0020414
          Stem-loop Sequence
          GCUUGAAGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUUCAUGACGAAUCAUACACGGUUGACCUAUUUUUCAGUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023786
          miRBase ID: cgr-miR-154-5p
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-154a
          Stem-loop Accession # MI0039004
          Stem-loop Sequence
          UAGGUUAUCCGUGUUGCCUUCGCUUUAUUUGUGCCGAAUCAUACACGGUUCACCUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047683
          miRBase ID: dno-miR-154a-5p
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-154
          Stem-loop Accession # MI0002598
          Stem-loop Sequence
          GUGGUACUUGAAGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUUUGUGACGAAUCAUACACGGUUGACCUAUUUUUCAGUACCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002297
          miRBase ID: ggo-miR-154
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-154
          Stem-loop Accession # MI0038663
          Stem-loop Sequence
          UAGGUUAUCCGUGUUGCCUUCGUUUUAUUUAUGUCGAAUCAUACGCGGUUGACCUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047036
          miRBase ID: cpo-miR-154-5p
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-154a
          Stem-loop Accession # MI0012879
          Stem-loop Sequence
          AGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUCUGUGACGAAUCAUACACGGUUCACCUAUUUUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013129
          miRBase ID: eca-miR-154a
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-154
          Stem-loop Accession # MI0002600
          Stem-loop Sequence
          GAGGUACUUGAAGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUUUGUGACGAAUCAUACACGGUUGACCUAUUUUUCAGUACCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002299
          miRBase ID: mne-miR-154
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-154
          Stem-loop Accession # MI0002601
          Stem-loop Sequence
          GUGGUACUUGAAGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUUUGUGACGAAUCAUACACGGUUGACCUAUUUUUCAGUACCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002300
          miRBase ID: ppa-miR-154
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-154
          Stem-loop Accession # MI0002597
          Stem-loop Sequence
          GUGGUACUUGAAGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUUUGUGACGAAUCAUACACGGUUGACCUAUUUUUCAGUACCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002296
          miRBase ID: ptr-miR-154
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-154
          Stem-loop Accession # MI0002599
          Stem-loop Sequence
          GUGGUACUUGAAGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUUUGUGACGAAUCAUACACGGUUGACCUAUUUUUCAGUACCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002298
          miRBase ID: ppy-miR-154
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-154
          Stem-loop Accession # MI0039313
          Stem-loop Sequence
          UAGGUUAUCCGUGUUGCCUUCGCUGUACUCGUGACGAAUCAUACACGGUUGGCCUAUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048271
          miRBase ID: ocu-miR-154-5p
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-154
          Stem-loop Accession # MI0007644
          Stem-loop Sequence
          GAGGUACUUGAAGAUAGGUUAUCCGUGUUGCCUUCGCUUUAUUUGUGACGAAUCAUACACGGUUGACCUAUUUUUCAGUACCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006215
          miRBase ID: mml-miR-154-5p
          Mature miRNA Sequence: UAGGUUAUCCGUGUUGCCUUCG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465319_pri
          TaqMan™ Pri-miRNA Assay : Mm03306393_pri
          TaqMan™ Pri-miRNA Assay : Hs03303277_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10086
          Ambion® Pre-miR™ miRNA Precursor : PM10086
          TaqMan™ MicroRNA Assay : 000477
          mirVana® miRNA inhibitor : MH10086
          TaqMan™ Advanced miRNA Assay : 477925_mir
          TaqMan™ Advanced miRNA Assay : rno480952_mir
          TaqMan™ Advanced miRNA Assay : mmu480952_mir


          miRBase Alias:

          Human: hsa-miR-154 (v17)
          Mouse: mmu-miR-154 (v17)
          Rat: rno-miR-154 (v18)
          Enterovirus CA55-1988: eca-miR-154 (v20)
          Macaca mulatta: mml-miR-154 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline