Search
Search

| Assay Name: | hsa-miR-154-5p |
|---|---|
| miRBase Accession Number: | MI0000480 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGGUUAUCCGUGUUGCCUUCG |
| Species: | Human, Mouse, Rat, Cricetulus griseus, Dasypus novemcinctus, Gorilla gorilla, Guinea pig, Horse, Macaca nemestrina, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Rabbit, Rhesus monkey |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | UAGGUUAUCCGUGUUGCCUUCG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-154-5p | MIMAT0000452 |
| Mouse | mmu-miR-154-5p | MIMAT0000164 |
| Rat | rno-miR-154-5p | MIMAT0000856 |
| Cricetulus griseus | cgr-miR-154-5p | MIMAT0023786 |
| Dasypus novemcinctus | dno-miR-154a-5p | MIMAT0047683 |
| Gorilla gorilla | ggo-miR-154 | MIMAT0002297 |
| View More Rows | ||
| Guinea pig | cpo-miR-154-5p | MIMAT0047036 |
| Horse | eca-miR-154a | MIMAT0013129 |
| Macaca nemestrina | mne-miR-154 | MIMAT0002299 |
| Pan paniscus | ppa-miR-154 | MIMAT0002300 |
| Pan troglodytes | ptr-miR-154 | MIMAT0002296 |
| Pongo pygmaeus | ppy-miR-154 | MIMAT0002298 |
| Rabbit | ocu-miR-154-5p | MIMAT0048271 |
| Rhesus monkey | mml-miR-154-5p | MIMAT0006215 |