Search
Search

| Assay Name: | rno-miR-496-3p |
|---|---|
| miRBase Accession Number: | MI0012622 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGUAUUACAUGGCCAAUCUCC |
| Species: | Rat, Cricetulus griseus |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGUAUUACAUGGCCAAUCUCC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-496-3p | MIMAT0012860 |
| Cricetulus griseus | cgr-miR-496 | MIMAT0023969 |