Search
Search

| Assay Name: | rno-miR-496* |
|---|---|
| miRBase Accession Number: | MI0012622 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGUUGUCCAUGGUGUGUUC |
| Species: | Rat |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AGGUUGUCCAUGGUGUGUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-496-5p | MIMAT0017372 |