Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10801
          Assay Name: hsa-miR-182-3p
          miRBase Accession Number: MI0000272
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 129770383 - 129770492 [-] on Build GRCh38
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Species: Human, Chrysemys picta, Dasypus novemcinctus, Fugu rubripes, Gadus morhua, Guinea pig, Ophiophagus hannah, Paralichthys olivaceus, Pteropus alecto, Rabbit, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10801
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000116, mir-182

          Mature miRNA Sequence:

          UGGUUCUAGACUUGCCAACUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-182-3p MIMAT0000260
          Chrysemys picta cpi-miR-182-3p MIMAT0037799
          Dasypus novemcinctus dno-miR-182-3p MIMAT0047700
          Fugu rubripes fru-miR-182 MIMAT0003093
          Gadus morhua gmo-miR-182-3p MIMAT0044266
          Guinea pig cpo-miR-182-3p MIMAT0047049
          View More Rows
          Ophiophagus hannah oha-miR-182-3p MIMAT0036765
          Paralichthys olivaceus pol-miR-182-3p MIMAT0025439
          Pteropus alecto pal-miR-182-3p MIMAT0040303
          Rabbit ocu-miR-182-3p MIMAT0048286
          Tetraodon nigroviridis tni-miR-182 MIMAT0003094
          Xenopus tropicalis xtr-miR-182-3p MIMAT0003611
          Zebrafish dre-miR-182-3p MIMAT0001272

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-182
          Stem-loop Accession # MI0000272
          Stem-loop Sequence
          GAGCUGCUUGCCUCCCCCCGUUUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAACAGGAUCCGGUGGUUCUAGACUUGCCAACUAUGGGGCGAGGACUCAGCCGGCAC
          Chromosome Location Chr. 7 - 129770383 - 129770492 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000260
          miRBase ID: hsa-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: Chr. 7 - 129770383 - 129770492 [-] on Build GRCh38

          Chrysemys picta

          Stem-loop ID cpi-mir-182
          Stem-loop Accession # MI0029506
          Stem-loop Sequence
          UCUCUCUCGCUGUCUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAUCAGGAUCCGGUGGUUCUAGACUUGCCAACUACCGCCCGAGGGUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037799
          miRBase ID: cpi-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-182
          Stem-loop Accession # MI0039013
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047700
          miRBase ID: dno-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-182
          Stem-loop Accession # MI0003457
          Stem-loop Sequence
          UGUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGAUGGAUCCGGUGGUUCUAGACUUGCCAACUACUGCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003093
          miRBase ID: fru-miR-182
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-182
          Stem-loop Accession # MI0036165
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGCUAGAAGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044266
          miRBase ID: gmo-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-182
          Stem-loop Accession # MI0038670
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047049
          miRBase ID: cpo-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-182
          Stem-loop Accession # MI0031379
          Stem-loop Sequence
          UGUCUUUGGCAAUGGUAGAACUCACACUGGUGAGCUAUCUGGUUCCGGUGGUUCUAGACUUGCCAACUACCGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036765
          miRBase ID: oha-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-mir-182
          Stem-loop Accession # MI0022202
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGAUGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025439
          miRBase ID: pol-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-182
          Stem-loop Accession # MI0032601
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCGCACUGGUGAGGUAAUGGGACCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040303
          miRBase ID: pal-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-182
          Stem-loop Accession # MI0039321
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAUGGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048286
          miRBase ID: ocu-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-182
          Stem-loop Accession # MI0003458
          Stem-loop Sequence
          UGUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGAUGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003094
          miRBase ID: tni-miR-182
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-182
          Stem-loop Accession # MI0004851
          Stem-loop Sequence
          AGUGGCUUCUCUGGCAGUGUUUGGCAAUGGUAGAACUCACACUGGUGAGCUAUGAAGAUCCGGUGGUUCUAGACUUGCCAACUAUGGCCUGGGAAUAAAGCAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003611
          miRBase ID: xtr-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-182
          Stem-loop Accession # MI0001368
          Stem-loop Sequence
          GUAUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGUCAGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001272
          miRBase ID: dre-miR-182-3p
          Mature miRNA Sequence: UGGUUCUAGACUUGCCAACUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302897_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10801
          TaqMan™ MicroRNA Assay : 000483
          mirVana® miRNA inhibitor : MH10801
          mirVana® miRNA mimic : MC10801
          TaqMan™ Advanced miRNA Assay : 478729_mir


          miRBase Alias:

          Human: hsa-miR-182* (v17)
          Danio rerio: dre-miR-182* (v18)
          Xenopus tropicalis: xtr-miR-182* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline