Search
Search

| Assay Name: | hsa-miR-182-3p |
|---|---|
| miRBase Accession Number: | MI0000272 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGUUCUAGACUUGCCAACUA |
| Species: | Human, Chrysemys picta, Dasypus novemcinctus, Fugu rubripes, Gadus morhua, Guinea pig, Ophiophagus hannah, Paralichthys olivaceus, Pteropus alecto, Rabbit, Tetraodon nigroviridis, Xenopus tropicalis, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000116, mir-182 |
Mature miRNA Sequence: | UGGUUCUAGACUUGCCAACUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-182-3p | MIMAT0000260 |
| Chrysemys picta | cpi-miR-182-3p | MIMAT0037799 |
| Dasypus novemcinctus | dno-miR-182-3p | MIMAT0047700 |
| Fugu rubripes | fru-miR-182 | MIMAT0003093 |
| Gadus morhua | gmo-miR-182-3p | MIMAT0044266 |
| Guinea pig | cpo-miR-182-3p | MIMAT0047049 |
| View More Rows | ||
| Ophiophagus hannah | oha-miR-182-3p | MIMAT0036765 |
| Paralichthys olivaceus | pol-miR-182-3p | MIMAT0025439 |
| Pteropus alecto | pal-miR-182-3p | MIMAT0040303 |
| Rabbit | ocu-miR-182-3p | MIMAT0048286 |
| Tetraodon nigroviridis | tni-miR-182 | MIMAT0003094 |
| Xenopus tropicalis | xtr-miR-182-3p | MIMAT0003611 |
| Zebrafish | dre-miR-182-3p | MIMAT0001272 |