Search
Search

| Assay Name: | hsa-miR-15b-5p |
|---|---|
| miRBase Accession Number: | MI0000438 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGCAGCACAUCAUGGUUUACA |
| Species: | Human, Mouse, Rat, Ateles geoffroyi, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Eptesicus fuscus, Goat, Gorilla gorilla, Horse, Lagothrix lagotricha, Macaca nemestrina, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000006, mir-15 |
Mature miRNA Sequence: | UAGCAGCACAUCAUGGUUUACA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-15b-5p | MIMAT0000417 |
| Mouse | mmu-miR-15b-5p | MIMAT0000124 |
| Rat | rno-miR-15b-5p | MIMAT0000784 |
| Ateles geoffroyi | age-miR-15b | MIMAT0002203 |
| Bovine | bta-miR-15b | MIMAT0003792 |
| Callithrix jacchus | cja-miR-15b | MIMAT0039439 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-15b-5p | MIMAT0023791 |
| Dasypus novemcinctus | dno-miR-15b-5p | MIMAT0047503 |
| Eptesicus fuscus | efu-miR-15 | MIMAT0035188 |
| Goat | chi-miR-15b-5p | MIMAT0035992 |
| Gorilla gorilla | ggo-miR-15b | MIMAT0002202 |
| Horse | eca-miR-15b | MIMAT0012954 |
| Lagothrix lagotricha | lla-miR-15b | MIMAT0002208 |
| Macaca nemestrina | mne-miR-15b | MIMAT0002209 |
| Nomascus leucogenys | nle-miR-15b | MIMAT0049460 |
| Otolemur garnettii | oga-miR-15b | MIMAT0049610 |
| Pan paniscus | ppa-miR-15b | MIMAT0002204 |
| Pan troglodytes | ptr-miR-15b | MIMAT0002206 |
| Pig | ssc-miR-15b | MIMAT0002125 |
| Pongo pygmaeus | ppy-miR-15b | MIMAT0002205 |
| Pteropus alecto | pal-miR-15b-5p | MIMAT0039905 |
| Rabbit | ocu-miR-15b-5p | MIMAT0048103 |
| Rhesus monkey | mml-miR-15b-5p | MIMAT0002207 |
| Tupaia chinensis | tch-miR-15b-5p | MIMAT0036552 |