Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10904
          Assay Name: hsa-miR-15b-5p
          miRBase Accession Number: MI0000438
          miRBase Version: v22.1
          Chromosome Location: Chr.3: 160404588 - 160404685 [+] on Build GRCh38
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Species: Human, Mouse, Rat, Ateles geoffroyi, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Eptesicus fuscus, Goat, Gorilla gorilla, Horse, Lagothrix lagotricha, Macaca nemestrina, Nomascus leucogenys, Otolemur garnettii, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis
          Product Type: mirVana® miRNA mimic
          Assay ID MC10904

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000006, mir-15

          Mature miRNA Sequence:

          UAGCAGCACAUCAUGGUUUACA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-15b-5p MIMAT0000417
          Mouse mmu-miR-15b-5p MIMAT0000124
          Rat rno-miR-15b-5p MIMAT0000784
          Ateles geoffroyi age-miR-15b MIMAT0002203
          Bovine bta-miR-15b MIMAT0003792
          Callithrix jacchus cja-miR-15b MIMAT0039439
          View More Rows
          Cricetulus griseus cgr-miR-15b-5p MIMAT0023791
          Dasypus novemcinctus dno-miR-15b-5p MIMAT0047503
          Eptesicus fuscus efu-miR-15 MIMAT0035188
          Goat chi-miR-15b-5p MIMAT0035992
          Gorilla gorilla ggo-miR-15b MIMAT0002202
          Horse eca-miR-15b MIMAT0012954
          Lagothrix lagotricha lla-miR-15b MIMAT0002208
          Macaca nemestrina mne-miR-15b MIMAT0002209
          Nomascus leucogenys nle-miR-15b MIMAT0049460
          Otolemur garnettii oga-miR-15b MIMAT0049610
          Pan paniscus ppa-miR-15b MIMAT0002204
          Pan troglodytes ptr-miR-15b MIMAT0002206
          Pig ssc-miR-15b MIMAT0002125
          Pongo pygmaeus ppy-miR-15b MIMAT0002205
          Pteropus alecto pal-miR-15b-5p MIMAT0039905
          Rabbit ocu-miR-15b-5p MIMAT0048103
          Rhesus monkey mml-miR-15b-5p MIMAT0002207
          Tupaia chinensis tch-miR-15b-5p MIMAT0036552

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-15b
          Stem-loop Accession # MI0000438
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUGCUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location Chr. 3 - 160404588 - 160404685 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000417
          miRBase ID: hsa-miR-15b-5p
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: Chr. 3 - 160404588 - 160404685 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-15b
          Stem-loop Accession # MI0000140
          Stem-loop Sequence
          CUGUAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAG
          Chromosome Location Chr. 3 - 69009772 - 69009835 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000124
          miRBase ID: mmu-miR-15b-5p
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: Chr. 3 - 69009772 - 69009835 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-15b
          Stem-loop Accession # MI0000843
          Stem-loop Sequence
          UUGGAACCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location Chr. 2 - 165605923 - 165606020 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000784
          miRBase ID: rno-miR-15b-5p
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: Chr. 2 - 165605923 - 165606020 [+] on Build Rnor_6.0

          Ateles geoffroyi

          Stem-loop ID age-mir-15b
          Stem-loop Accession # MI0002492
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002203
          miRBase ID: age-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-15b
          Stem-loop Accession # MI0005012
          Stem-loop Sequence
          UUGAGACCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003792
          miRBase ID: bta-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-15b
          Stem-loop Accession # MI0031961
          Stem-loop Sequence
          UGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039439
          miRBase ID: cja-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-15b
          Stem-loop Accession # MI0020417
          Stem-loop Sequence
          AAAGUACUAUAGCAGCACAUCAUGGUUUACAUACUAUAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023791
          miRBase ID: cgr-miR-15b-5p
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-15b
          Stem-loop Accession # MI0038906
          Stem-loop Sequence
          UAGCAGCACAUCAUGGUUUACAUACUGCAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047503
          miRBase ID: dno-miR-15b-5p
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-15
          Stem-loop Accession # MI0028900
          Stem-loop Sequence
          UAAAGUACUGUAGCAGCACAUCAUGGUUUACAUACUAUAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035188
          miRBase ID: efu-miR-15
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-15b
          Stem-loop Accession # MI0030645
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035992
          miRBase ID: chi-miR-15b-5p
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-15b
          Stem-loop Accession # MI0002491
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUGCUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002202
          miRBase ID: ggo-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-15b
          Stem-loop Accession # MI0012706
          Stem-loop Sequence
          CUGUAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012954
          miRBase ID: eca-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Lagothrix lagotricha

          Stem-loop ID lla-mir-15b
          Stem-loop Accession # MI0002497
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002208
          miRBase ID: lla-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-15b
          Stem-loop Accession # MI0002498
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002209
          miRBase ID: mne-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-15b
          Stem-loop Accession # MI0040214
          Stem-loop Sequence
          UAGCAGCACAUCAUGGUUUACAUGCUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049460
          miRBase ID: nle-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-15b
          Stem-loop Accession # MI0040391
          Stem-loop Sequence
          UAGCAGCACAUCAUGGUUUACAUACUGCAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049610
          miRBase ID: oga-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-15b
          Stem-loop Accession # MI0002493
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUGCUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002204
          miRBase ID: ppa-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-15b
          Stem-loop Accession # MI0002495
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUGCUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002206
          miRBase ID: ptr-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-15b
          Stem-loop Accession # MI0002419
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUACUACAAUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002125
          miRBase ID: ssc-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-15b
          Stem-loop Accession # MI0002494
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUGCUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002205
          miRBase ID: ppy-miR-15b
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-15b
          Stem-loop Accession # MI0032393
          Stem-loop Sequence
          UAGCAGCACAUCAUGGUUUACAUACUACAGUCGAUGCGAAUCAUUAUUUGCUGCUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039905
          miRBase ID: pal-miR-15b-5p
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-15b
          Stem-loop Accession # MI0039222
          Stem-loop Sequence
          UAGCAGCACAUCAUGGUUUACAUAAUAUAGUCAAGAUGCGAAUCAUAAUUUGCUGCUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048103
          miRBase ID: ocu-miR-15b-5p
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-15b
          Stem-loop Accession # MI0002496
          Stem-loop Sequence
          UUGAGGCCUUAAAGUACUGUAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUAGAAAUUUAAGGAAAUUCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002207
          miRBase ID: mml-miR-15b-5p
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-15b
          Stem-loop Accession # MI0031212
          Stem-loop Sequence
          UAGCAGCACAUCAUGGUUUACAUACUACAGUCAAGAUGCGAAUCAUUAUUUGCUGCUCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036552
          miRBase ID: tch-miR-15b-5p
          Mature miRNA Sequence: UAGCAGCACAUCAUGGUUUACA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306803_pri
          TaqMan™ Pri-miRNA Assay : Rn03464523_pri
          TaqMan™ Pri-miRNA Assay : Hs03294999_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10904
          Ambion® Pre-miR™ miRNA Precursor : PM10904
          TaqMan™ MicroRNA Assay : 000390
          mirVana® miRNA inhibitor : MH10904
          TaqMan™ Advanced miRNA Assay : mmu482957_mir
          TaqMan™ Advanced miRNA Assay : rno481454_mir
          TaqMan™ Advanced miRNA Assay : 478313_mir


          miRBase Alias:

          Human: hsa-miR-15b (v17)
          Mouse: mmu-miR-15b (v17)
          Rat: rno-miR-15b (v18)
          Macaca mulatta: mml-miR-15b (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline