Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM10951
          Assay Name: hsa-miR-93-5p
          miRBase Accession Number: MI0000095
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 100093768 - 100093847 [-] on Build GRCh38
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Goat, Guinea pig, Horse, Nomascus leucogenys, Otolemur garnettii, Papio hamadryas, Pteropus alecto, Rabbit, Tupaia chinensis, Xenopus laevis
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM10951
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          CAAAGUGCUGUUCGUGCAGGUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-93-5p MIMAT0000093
          Mouse mmu-miR-93-5p MIMAT0000540
          Rat rno-miR-93-5p MIMAT0000817
          Alligator mississippiensis ami-miR-93-5p MIMAT0038104
          Callithrix jacchus cja-miR-93 MIMAT0039390
          Cricetulus griseus cgr-miR-93-5p MIMAT0024013
          View More Rows
          Dasypus novemcinctus dno-miR-93-5p MIMAT0047586
          Daubentonia madagascariensis dma-miR-93 MIMAT0049285
          Dog cfa-miR-93 MIMAT0006696
          Goat chi-miR-93-5p MIMAT0036305
          Guinea pig cpo-miR-93-5p MIMAT0046933
          Horse eca-miR-93 MIMAT0013060
          Nomascus leucogenys nle-miR-93 MIMAT0049351
          Otolemur garnettii oga-miR-93 MIMAT0049702
          Papio hamadryas pha-miR-93 MIMAT0049552
          Pteropus alecto pal-miR-93-5p MIMAT0039937
          Rabbit ocu-miR-93-5p MIMAT0048176
          Tupaia chinensis tch-miR-93-5p MIMAT0036617
          Xenopus laevis xla-miR-93-5p MIMAT0046613

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-93
          Stem-loop Accession # MI0000095
          Stem-loop Sequence
          CUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCCAACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCGG
          Chromosome Location Chr. 7 - 100093768 - 100093847 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000093
          miRBase ID: hsa-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: Chr. 7 - 100093768 - 100093847 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-93
          Stem-loop Accession # MI0000581
          Stem-loop Sequence
          AGUCAUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUAAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCAGGACA
          Chromosome Location Chr. 5 - 138165523 - 138165610 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000540
          miRBase ID: mmu-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: Chr. 5 - 138165523 - 138165610 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-93
          Stem-loop Accession # MI0000880
          Stem-loop Sequence
          AGUCAUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGCAUUGCCUGACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCAGGACA
          Chromosome Location Chr. 12 - 19307543 - 19307629 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000817
          miRBase ID: rno-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: Chr. 12 - 19307543 - 19307629 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-93
          Stem-loop Accession # MI0029705
          Stem-loop Sequence
          CUUGAGCUGCGGUGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUGUGCAGCCAUCUACUGCCGGGCAGCACUUCCCGAGCCCCCGCAGCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038104
          miRBase ID: ami-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-93
          Stem-loop Accession # MI0031909
          Stem-loop Sequence
          GUCCUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCCAACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCGGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039390
          miRBase ID: cja-miR-93
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-93
          Stem-loop Accession # MI0020562
          Stem-loop Sequence
          AUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUAAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGAGCCCCCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024013
          miRBase ID: cgr-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-93
          Stem-loop Accession # MI0038950
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047586
          miRBase ID: dno-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Daubentonia madagascariensis

          Stem-loop ID dma-mir-93
          Stem-loop Accession # MI0040016
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049285
          miRBase ID: dma-miR-93
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-93
          Stem-loop Accession # MI0008110
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006696
          miRBase ID: cfa-miR-93
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-93
          Stem-loop Accession # MI0030841
          Stem-loop Sequence
          CUGGGGGCUCCAAAGUGCUGUUCGUGCAGGUAGUGUGAUCACCUGACCUACUGCUGAGCCAGCACUUCCCGAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036305
          miRBase ID: chi-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-93
          Stem-loop Accession # MI0038606
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046933
          miRBase ID: cpo-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-93
          Stem-loop Accession # MI0012804
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013060
          miRBase ID: eca-miR-93
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-93
          Stem-loop Accession # MI0040088
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCCGACCUACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049351
          miRBase ID: nle-miR-93
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-93
          Stem-loop Accession # MI0040486
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049702
          miRBase ID: oga-miR-93
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-93
          Stem-loop Accession # MI0040320
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUAACCGACCUACUGCUGAGCUAGCACUUCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049552
          miRBase ID: pha-miR-93
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-93
          Stem-loop Accession # MI0032409
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039937
          miRBase ID: pal-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-93
          Stem-loop Accession # MI0039261
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048176
          miRBase ID: ocu-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-93
          Stem-loop Accession # MI0031290
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGUGUGAUUACCUGACCUACUGCUGAGCUAGCACUUCCCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036617
          miRBase ID: tch-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-93
          Stem-loop Accession # MI0038341
          Stem-loop Sequence
          CAAAGUGCUGUUCGUGCAGGUAGCUUUAUAACAGACCUACUGCAUGAGCAGCACUUCCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046613
          miRBase ID: xla-miR-93-5p
          Mature miRNA Sequence: CAAAGUGCUGUUCGUGCAGGUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm04227694_pri
          TaqMan™ Pri-miRNA Assay : Rn03465019_pri
          TaqMan™ Pri-miRNA Assay : Hs04227110_pri
          Ambion® Pre-miR™ miRNA Precursor : PM10951
          TaqMan™ MicroRNA Assay : 001090
          mirVana® miRNA inhibitor : MH10951
          mirVana® miRNA mimic : MC10951
          TaqMan™ Advanced miRNA Assay : mmu481280_mir
          TaqMan™ Advanced miRNA Assay : rno481280_mir
          TaqMan™ Advanced miRNA Assay : 478210_mir


          miRBase Alias:

          Human: hsa-miR-93 (v17)
          Mouse: mmu-miR-93 (v17)
          Rat: rno-miR-93 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline