Search
Search

| Assay Name: | rno-miR-93-5p |
|---|---|
| Stem-loop Accession Number: | MI0000095 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAAGUGCUGUUCGUGCAGGUAG |
| Species: | Human, Mouse, Rat, Alligator mississippiensis, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Daubentonia madagascariensis, Dog, Goat, Guinea pig, Horse, Nomascus leucogenys, Otolemur garnettii, Papio hamadryas, Pteropus alecto, Rabbit, Tupaia chinensis, Xenopus laevis |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | CAAAGUGCUGUUCGUGCAGGUAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-93-5p | MIMAT0000093 |
| Mouse | mmu-miR-93-5p | MIMAT0000540 |
| Rat | rno-miR-93-5p | MIMAT0000817 |
| Alligator mississippiensis | ami-miR-93-5p | MIMAT0038104 |
| Callithrix jacchus | cja-miR-93 | MIMAT0039390 |
| Cricetulus griseus | cgr-miR-93-5p | MIMAT0024013 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-93-5p | MIMAT0047586 |
| Daubentonia madagascariensis | dma-miR-93 | MIMAT0049285 |
| Dog | cfa-miR-93 | MIMAT0006696 |
| Goat | chi-miR-93-5p | MIMAT0036305 |
| Guinea pig | cpo-miR-93-5p | MIMAT0046933 |
| Horse | eca-miR-93 | MIMAT0013060 |
| Nomascus leucogenys | nle-miR-93 | MIMAT0049351 |
| Otolemur garnettii | oga-miR-93 | MIMAT0049702 |
| Papio hamadryas | pha-miR-93 | MIMAT0049552 |
| Pteropus alecto | pal-miR-93-5p | MIMAT0039937 |
| Rabbit | ocu-miR-93-5p | MIMAT0048176 |
| Tupaia chinensis | tch-miR-93-5p | MIMAT0036617 |
| Xenopus laevis | xla-miR-93-5p | MIMAT0046613 |