Search
Search

| Assay Name: | |
|---|---|
| miRBase Accession Number: | MI0009783 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAAGUCACUAGUGGUUCCGUUUA |
| Species: | Bovine, Gorilla gorilla, Macaca nemestrina, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000088, mir-224 |
Mature miRNA Sequence: | CAAGUCACUAGUGGUUCCGUUUA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-224 | MIMAT0009271 |
| Gorilla gorilla | ggo-miR-224 | MIMAT0002596 |
| Macaca nemestrina | mne-miR-224 | MIMAT0002598 |
| Pan paniscus | ppa-miR-224 | MIMAT0002599 |
| Pan troglodytes | ptr-miR-224 | MIMAT0002595 |
| Pig | ssc-miR-224 | MIMAT0002132 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-224 | MIMAT0002597 |
| Rhesus monkey | mml-miR-224-5p | MIMAT0002594 |