Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11010
          Assay Name:
          miRBase Accession Number: MI0009783
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: CAAGUCACUAGUGGUUCCGUUUA
          Species: Bovine, Gorilla gorilla, Macaca nemestrina, Pan paniscus, Pan troglodytes, Pig, Pongo pygmaeus, Rhesus monkey
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11010
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000088, mir-224

          Mature miRNA Sequence:

          CAAGUCACUAGUGGUUCCGUUUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Bovine bta-miR-224 MIMAT0009271
          Gorilla gorilla ggo-miR-224 MIMAT0002596
          Macaca nemestrina mne-miR-224 MIMAT0002598
          Pan paniscus ppa-miR-224 MIMAT0002599
          Pan troglodytes ptr-miR-224 MIMAT0002595
          Pig ssc-miR-224 MIMAT0002132
          View More Rows
          Pongo pygmaeus ppy-miR-224 MIMAT0002597
          Rhesus monkey mml-miR-224-5p MIMAT0002594

          Stem-loop Details



          Bovine

          Stem-loop ID bta-mir-224
          Stem-loop Accession # MI0009783
          Stem-loop Sequence
          GGGCUUUCAAGUCACUAGUGGUUCCGUUUAGUAGAUGGUUGUACAUUGUUUCAAAAUGGUACCCUAGUGACUACAAAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009271
          miRBase ID: bta-miR-224
          Mature miRNA Sequence: CAAGUCACUAGUGGUUCCGUUUA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-224
          Stem-loop Accession # MI0002905
          Stem-loop Sequence
          GGGCUUUCAAGUCACUAGUGGUUCCGUUUAGUAGAUGAUUGUGCAUUGUUUCAAAAUGGUGCCCUAGUGACUACAAAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002596
          miRBase ID: ggo-miR-224
          Mature miRNA Sequence: CAAGUCACUAGUGGUUCCGUUUA
          Chromosome Location: NA

          Macaca nemestrina

          Stem-loop ID mne-mir-224
          Stem-loop Accession # MI0002907
          Stem-loop Sequence
          GGGCUUUCAAGUCACUAGUGGUUCCGUUUAGUAGAUGAUUGUGCAUUGUUUCAAAAUGGUGCCCUAGUGACUACAAAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002598
          miRBase ID: mne-miR-224
          Mature miRNA Sequence: CAAGUCACUAGUGGUUCCGUUUA
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-224
          Stem-loop Accession # MI0002908
          Stem-loop Sequence
          GGGCUUUCAAGUCACUAGUGGUUCCGUUUAGUAGAUGAUUGUGCAUUGUUUCAAAAUGGUGCCCUAGUGACUACAAAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002599
          miRBase ID: ppa-miR-224
          Mature miRNA Sequence: CAAGUCACUAGUGGUUCCGUUUA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-224
          Stem-loop Accession # MI0002904
          Stem-loop Sequence
          GGGCUUUCAAGUCACUAGUGGUUCCGUUUAGUAGAUGAUUGUGCAUUGUUUCAAAAUGGUGCCCUAGUGACUACAAAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002595
          miRBase ID: ptr-miR-224
          Mature miRNA Sequence: CAAGUCACUAGUGGUUCCGUUUA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-224
          Stem-loop Accession # MI0002426
          Stem-loop Sequence
          GGGCUUUCAAGUCACUAGUGGUUCCGUUUAGUAGAAGGUUUUGCAUUGUUUCAAAAUGGUGCCCUAGUGACUACAAAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002132
          miRBase ID: ssc-miR-224
          Mature miRNA Sequence: CAAGUCACUAGUGGUUCCGUUUA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-224
          Stem-loop Accession # MI0002906
          Stem-loop Sequence
          GGGCUUUCAAGUCACUAGUGGUUCCGUUUAGUAGAUGAUUGUGCAUUGUUUCAAAAUGGUGCCCUAGUGACUACAAAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002597
          miRBase ID: ppy-miR-224
          Mature miRNA Sequence: CAAGUCACUAGUGGUUCCGUUUA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-224
          Stem-loop Accession # MI0002903
          Stem-loop Sequence
          GGGCUUUCAAGUCACUAGUGGUUCCGUUUAGUAGAUGAUUGUGCAUUGUUUCAAAAUGGUGCCCUAGUGACUACAAAGCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002594
          miRBase ID: mml-miR-224-5p
          Mature miRNA Sequence: CAAGUCACUAGUGGUUCCGUUUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11010
          TaqMan™ MicroRNA Assay : 000599
          mirVana® miRNA inhibitor : MH11010
          mirVana® miRNA mimic : MC11010


          miRBase Alias:

          Human: hsa-miR-224 (v9.2)
          Rat: rno-miR-224 (v15)
          Macaca mulatta: mml-miR-224 (v19)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline