Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM11090
          Assay Name:
          miRBase Accession Number: MI0033720
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Species: Anolis carolinensis, Astatotilapia burtoni, Callithrix jacchus, Gadus morhua, Gorilla gorilla, Ictalurus punctatus, Metriaclima zebra, Monodelphis domestica, Neolamprologus brichardi, Oreochromis niloticus, Ornithorhynchus anatinus, Paralichthys olivaceus, Pongo pygmaeus, Pundamilia nyererei, Rhesus monkey, Salmo salar, Xenopus tropicalis, Zebrafish
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM11090
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Mature miRNA Sequence:

          UUUGGCAAUGGUAGAACUCACA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Anolis carolinensis aca-miR-182-5p MIMAT0021811
          Astatotilapia burtoni abu-miR-182 MIMAT0042044
          Callithrix jacchus cja-miR-182 MIMAT0039533
          Gadus morhua gmo-miR-182-5p MIMAT0044265
          Gorilla gorilla ggo-miR-182 MIMAT0024298
          Ictalurus punctatus ipu-miR-182 MIMAT0029439
          View More Rows
          Metriaclima zebra mze-miR-182 MIMAT0042280
          Monodelphis domestica mdo-miR-182-5p MIMAT0004118
          Neolamprologus brichardi nbr-miR-182 MIMAT0042469
          Oreochromis niloticus oni-miR-182 MIMAT0042646
          Ornithorhynchus anatinus oan-miR-182-5p MIMAT0012798
          Paralichthys olivaceus pol-miR-182-5p MIMAT0025438
          Pongo pygmaeus ppy-miR-182 MIMAT0002514
          Pundamilia nyererei pny-miR-182 MIMAT0042856
          Rhesus monkey mml-miR-182 MIMAT0002513
          Salmo salar ssa-miR-182-5p MIMAT0032427
          Xenopus tropicalis xtr-miR-182-5p MIMAT0003610
          Zebrafish dre-miR-182-5p MIMAT0001271

          Stem-loop Details



          Anolis carolinensis

          Stem-loop ID aca-mir-182
          Stem-loop Accession # MI0018780
          Stem-loop Sequence
          GUUUUCUCCGGCUGUUUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAUCAGGAUCCGGUGGUUCUAGACUUGCCAACUACGGCCCGAGGGUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021811
          miRBase ID: aca-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-mir-182
          Stem-loop Accession # MI0033720
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGAUGGAUCCGGUGGUUCUAGACUUGCCAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042044
          miRBase ID: abu-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-182
          Stem-loop Accession # MI0032063
          Stem-loop Sequence
          CCUCUCCCCGUUUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUAUGGGGCGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039533
          miRBase ID: cja-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Gadus morhua

          Stem-loop ID gmo-mir-182
          Stem-loop Accession # MI0036165
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGCUAGAAGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0044265
          miRBase ID: gmo-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-182
          Stem-loop Accession # MI0020846
          Stem-loop Sequence
          GCUGCUUGCCUCCCCCUGUUUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAACAGGAUCCGGUGGUUCUAGACUUGCCAACUAUGGGGCGAGGACUCAGCCGGCACCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024298
          miRBase ID: ggo-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-182
          Stem-loop Accession # MI0024534
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGUCAGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029439
          miRBase ID: ipu-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Metriaclima zebra

          Stem-loop ID mze-mir-182
          Stem-loop Accession # MI0034016
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGAUGGAUCCGGUGGUUCUAGACUUGCCAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042280
          miRBase ID: mze-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-182
          Stem-loop Accession # MI0005307
          Stem-loop Sequence
          GCUGUGUUUGGCAAUGGUAGAACUCACACUGGUGAGAUAAUGGAAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004118
          miRBase ID: mdo-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Neolamprologus brichardi

          Stem-loop ID nbr-mir-182
          Stem-loop Accession # MI0034278
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGAUGGAUCCGGUGGUUCUAGACUUGCCAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042469
          miRBase ID: nbr-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-182
          Stem-loop Accession # MI0034524
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGAUGGAUCCGGUGGUUCUAGACUUGCCAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042646
          miRBase ID: oni-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-182
          Stem-loop Accession # MI0012558
          Stem-loop Sequence
          CCCUCCUGCUGUGUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUAUGGCCCGAGGGCUCAGCCGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0012798
          miRBase ID: oan-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-mir-182
          Stem-loop Accession # MI0022202
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGAUGGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025438
          miRBase ID: pol-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-182
          Stem-loop Accession # MI0002816
          Stem-loop Sequence
          CACAGCUGCUUGCCUUCCCCCGUUUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAACAGGAUCCGGUGGUUCUAGACUUGCCAACUAUGGGGCGAGGACUCAGCCGGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002514
          miRBase ID: ppy-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Pundamilia nyererei

          Stem-loop ID pny-mir-182
          Stem-loop Accession # MI0034796
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGAUGGAUCCGGUGGUUCUAGACUUGCCAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042856
          miRBase ID: pny-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-182
          Stem-loop Accession # MI0002815
          Stem-loop Sequence
          CACAGCUGCUUGCCUCCCCCUGUUUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAAUGGGAUCCGGUGGUUCUAGACUUGCCAACUACGGGGCGAGGGCUCAGCCGGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002513
          miRBase ID: mml-miR-182
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-182
          Stem-loop Accession # MI0026543
          Stem-loop Sequence
          UUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGCUGGAUCCGGUGGUUCUAGACUUGCCAACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032427
          miRBase ID: ssa-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-182
          Stem-loop Accession # MI0004851
          Stem-loop Sequence
          AGUGGCUUCUCUGGCAGUGUUUGGCAAUGGUAGAACUCACACUGGUGAGCUAUGAAGAUCCGGUGGUUCUAGACUUGCCAACUAUGGCCUGGGAAUAAAGCAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003610
          miRBase ID: xtr-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-182
          Stem-loop Accession # MI0001368
          Stem-loop Sequence
          GUAUUUGGCAAUGGUAGAACUCACACUGGUGAGGUAGUCAGAUCCGGUGGUUCUAGACUUGCCAACUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001271
          miRBase ID: dre-miR-182-5p
          Mature miRNA Sequence: UUUGGCAAUGGUAGAACUCACA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          Ambion® Pre-miR™ miRNA Precursor : PM11090
          TaqMan™ MicroRNA Assay : 000597
          mirVana® miRNA inhibitor : MH11090
          mirVana® miRNA mimic : MC11090


          miRBase Alias:

          Human: hsa-miR-182 (v9.2)
          Mouse: mmu-miR-182 (v9.2)
          Aeromonas caviae Ae398: aca-miR-182 (v18)
          Danio rerio: dre-miR-182 (v18)
          Monodelphis domestica: mdo-miR-182 (v19)
          Ochrobactrum anthropi: oan-miR-182 (v19)
          Xenopus tropicalis: xtr-miR-182 (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline