Search
Search

| Assay Name: | |
|---|---|
| miRBase Accession Number: | MI0033720 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUGGCAAUGGUAGAACUCACA |
| Species: | Anolis carolinensis, Astatotilapia burtoni, Callithrix jacchus, Gadus morhua, Gorilla gorilla, Ictalurus punctatus, Metriaclima zebra, Monodelphis domestica, Neolamprologus brichardi, Oreochromis niloticus, Ornithorhynchus anatinus, Paralichthys olivaceus, Pongo pygmaeus, Pundamilia nyererei, Rhesus monkey, Salmo salar, Xenopus tropicalis, Zebrafish |
| Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | UUUGGCAAUGGUAGAACUCACA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-182-5p | MIMAT0021811 |
| Astatotilapia burtoni | abu-miR-182 | MIMAT0042044 |
| Callithrix jacchus | cja-miR-182 | MIMAT0039533 |
| Gadus morhua | gmo-miR-182-5p | MIMAT0044265 |
| Gorilla gorilla | ggo-miR-182 | MIMAT0024298 |
| Ictalurus punctatus | ipu-miR-182 | MIMAT0029439 |
| View More Rows | ||
| Metriaclima zebra | mze-miR-182 | MIMAT0042280 |
| Monodelphis domestica | mdo-miR-182-5p | MIMAT0004118 |
| Neolamprologus brichardi | nbr-miR-182 | MIMAT0042469 |
| Oreochromis niloticus | oni-miR-182 | MIMAT0042646 |
| Ornithorhynchus anatinus | oan-miR-182-5p | MIMAT0012798 |
| Paralichthys olivaceus | pol-miR-182-5p | MIMAT0025438 |
| Pongo pygmaeus | ppy-miR-182 | MIMAT0002514 |
| Pundamilia nyererei | pny-miR-182 | MIMAT0042856 |
| Rhesus monkey | mml-miR-182 | MIMAT0002513 |
| Salmo salar | ssa-miR-182-5p | MIMAT0032427 |
| Xenopus tropicalis | xtr-miR-182-5p | MIMAT0003610 |
| Zebrafish | dre-miR-182-5p | MIMAT0001271 |