Search
Search

| Assay Name: | hsa-miR-491-5p |
|---|---|
| miRBase Accession Number: | MI0003126 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGUGGGGAACCCUUCCAUGAGG |
| Species: | Human, Mouse, Bovine, Callithrix jacchus, Goat, Horse, Pig, Pongo pygmaeus, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000319, mir-491 |
Mature miRNA Sequence: | AGUGGGGAACCCUUCCAUGAGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-491-5p | MIMAT0002807 |
| Mouse | mmu-miR-491-5p | MIMAT0003486 |
| Bovine | bta-miR-491 | MIMAT0009332 |
| Callithrix jacchus | cja-miR-491 | MIMAT0039375 |
| Goat | chi-miR-491-5p | MIMAT0036251 |
| Horse | eca-miR-491-5p | MIMAT0013116 |
| View More Rows | ||
| Pig | ssc-miR-491 | MIMAT0020592 |
| Pongo pygmaeus | ppy-miR-491-5p | MIMAT0015911 |
| Rhesus monkey | mml-miR-491-5p | MIMAT0006352 |