Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11479
          Assay Name: hsa-miR-491-5p
          miRBase Accession Number: MI0003126
          miRBase Version: v22.1
          Chromosome Location: Chr.9: 20716105 - 20716188 [+] on Build GRCh38
          Mature miRNA Sequence: AGUGGGGAACCCUUCCAUGAGG
          Species: Human, Mouse, Bovine, Callithrix jacchus, Goat, Horse, Pig, Pongo pygmaeus, Rhesus monkey
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11479
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000319, mir-491

          Mature miRNA Sequence:

          AGUGGGGAACCCUUCCAUGAGG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-491-5p MIMAT0002807
          Mouse mmu-miR-491-5p MIMAT0003486
          Bovine bta-miR-491 MIMAT0009332
          Callithrix jacchus cja-miR-491 MIMAT0039375
          Goat chi-miR-491-5p MIMAT0036251
          Horse eca-miR-491-5p MIMAT0013116
          View More Rows
          Pig ssc-miR-491 MIMAT0020592
          Pongo pygmaeus ppy-miR-491-5p MIMAT0015911
          Rhesus monkey mml-miR-491-5p MIMAT0006352

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-491
          Stem-loop Accession # MI0003126
          Stem-loop Sequence
          UUGACUUAGCUGGGUAGUGGGGAACCCUUCCAUGAGGAGUAGAACACUCCUUAUGCAAGAUUCCCUUCUACCUGGCUGGGUUGG
          Chromosome Location Chr. 9 - 20716105 - 20716188 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002807
          miRBase ID: hsa-miR-491-5p
          Mature miRNA Sequence: AGUGGGGAACCCUUCCAUGAGG
          Chromosome Location: Chr. 9 - 20716105 - 20716188 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-491
          Stem-loop Accession # MI0004680
          Stem-loop Sequence
          AAUUGACUUAGCUGGGAAGUGGGGAACCCUUCCAUGAGGAGUAGAACACUCCUUAUGCAAGAUUCCCUUCUACCUGACUGAGUUGA
          Chromosome Location Chr. 4 - 88122040 - 88122125 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003486
          miRBase ID: mmu-miR-491-5p
          Mature miRNA Sequence: AGUGGGGAACCCUUCCAUGAGG
          Chromosome Location: Chr. 4 - 88122040 - 88122125 [+] on Build GRCm38

          Bovine

          Stem-loop ID bta-mir-491
          Stem-loop Accession # MI0009846
          Stem-loop Sequence
          UUGACUUAGCUGGGUAGUGGGGAACCCUUCCAUGAGGAGUAGAACACUCCUUAUGCAAGAUUCCCUUCUACCUGACUGGGUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009332
          miRBase ID: bta-miR-491
          Mature miRNA Sequence: AGUGGGGAACCCUUCCAUGAGG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-491
          Stem-loop Accession # MI0031893
          Stem-loop Sequence
          GGAAUUGACUUAGCUGGGUAGUGGGGAACCCUUCCAUGAGGAGUAGAACACUCCUUAUGCAAGAUUCCCUUCUACCUGGCUGGGUUGGAGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039375
          miRBase ID: cja-miR-491
          Mature miRNA Sequence: AGUGGGGAACCCUUCCAUGAGG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-491
          Stem-loop Accession # MI0030806
          Stem-loop Sequence
          GGGAAUUGACUUAGCUGGGUAGUGGGGAACCCUUCCAUGAGGAGUAGAACACUCCUUAUGCAAGAUUCCCUUCUACCUGACUGGGUUGGAGUCAUAUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036251
          miRBase ID: chi-miR-491-5p
          Mature miRNA Sequence: AGUGGGGAACCCUUCCAUGAGG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-491
          Stem-loop Accession # MI0012867
          Stem-loop Sequence
          UUGACUUAGCUGGGUAGUGGGGAACCCUUCCAUGAGGAGUAGAACACUCCUUAUGCAAGAUUCCCUUCUACCUGACUGGGUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013116
          miRBase ID: eca-miR-491-5p
          Mature miRNA Sequence: AGUGGGGAACCCUUCCAUGAGG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-491
          Stem-loop Accession # MI0017993
          Stem-loop Sequence
          UUAGCUGGGUAGUGGGGAACCCUUCCAUGAGGAGUAGAACACUCCUUAUGCAAGAUUCCCUUCUACCUGACUGGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0020592
          miRBase ID: ssc-miR-491
          Mature miRNA Sequence: AGUGGGGAACCCUUCCAUGAGG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-491
          Stem-loop Accession # MI0014964
          Stem-loop Sequence
          UUGACUUAGCUGGGUAGUGGGGAACCCUUCCAUGAGGAGUAGAACACUCCUUAUGCAAGAUUCCCUUCUACCUGGCUGGGUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015911
          miRBase ID: ppy-miR-491-5p
          Mature miRNA Sequence: AGUGGGGAACCCUUCCAUGAGG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-491
          Stem-loop Accession # MI0007765
          Stem-loop Sequence
          UUGACUUAGCUGGGUAGUGGGGAACCCUUCCAUGAGGAGUAGAACACUCCUUAUGCAAGAUUCCCUUCUACCUGGCUGGGUUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006352
          miRBase ID: mml-miR-491-5p
          Mature miRNA Sequence: AGUGGGGAACCCUUCCAUGAGG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03308184_pri
          TaqMan™ Pri-miRNA Assay : Hs03303840_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11479
          TaqMan™ MicroRNA Assay : 001630
          mirVana® miRNA inhibitor : MH11479
          mirVana® miRNA mimic : MC11479
          TaqMan™ Advanced miRNA Assay : mmu481859_mir
          TaqMan™ Advanced miRNA Assay : 478132_mir


          miRBase Alias:

          Mouse: mmu-miR-491 (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline