Search
Search

| Assay Name: | hsa-miR-668-3p |
|---|---|
| miRBase Accession Number: | MI0003761 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUCACUCGGCUCGGCCCACUAC |
| Species: | Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000357, mir-668 |
Mature miRNA Sequence: | UGUCACUCGGCUCGGCCCACUAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-668-3p | MIMAT0003881 |
| Pan troglodytes | ptr-miR-668 | MIMAT0008313 |
| Pongo pygmaeus | ppy-miR-668 | MIMAT0016095 |
| Rhesus monkey | mml-miR-668 | MIMAT0006504 |