Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › 479151_mir

          Specificity testing was performed using human anti-targets.

          Assay Name: hsa-miR-668-3p
          Stem-loop Accession Number: MI0003761
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 101055258 - 101055323 [+] on Build GRCh38
          Mature miRNA Sequence: UGUCACUCGGCUCGGCCCACUAC
          Species: Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID 479151_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000357, mir-668

          Mature miRNA Sequence:

          UGUCACUCGGCUCGGCCCACUAC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-668-3p MIMAT0003881
          Pan troglodytes ptr-miR-668 MIMAT0008313
          Pongo pygmaeus ppy-miR-668 MIMAT0016095
          Rhesus monkey mml-miR-668 MIMAT0006504

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-668
          Stem-loop Accession # MI0003761
          Stem-loop Sequence
          GGUAAGUGCGCCUCGGGUGAGCAUGCACUUAAUGUGGGUGUAUGUCACUCGGCUCGGCCCACUACC
          Chromosome Location Chr. 14 - 101055258 - 101055323 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003881
          miRBase ID: hsa-miR-668-3p
          Mature miRNA Sequence: UGUCACUCGGCUCGGCCCACUAC
          Chromosome Location: Chr. 14 - 101055258 - 101055323 [+] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-668
          Stem-loop Accession # MI0008850
          Stem-loop Sequence
          GUAAGUGCGCCUCGGGUGAGCAUGCACUUAAUGUGGGUGUAUGUCACUCGGCUCGGCCCACUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008313
          miRBase ID: ptr-miR-668
          Mature miRNA Sequence: UGUCACUCGGCUCGGCCCACUAC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-668
          Stem-loop Accession # MI0015136
          Stem-loop Sequence
          GGUAAGUGCGCCUCGGGUGAGCAUGCACUUAAUGUGAGGUGUAUGUCACUCGGCUCGGCCCACUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0016095
          miRBase ID: ppy-miR-668
          Mature miRNA Sequence: UGUCACUCGGCUCGGCCCACUAC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-668
          Stem-loop Accession # MI0007906
          Stem-loop Sequence
          GGUAAGUGCGCCUCGGGUGAGCAUGCACUUAAUGUGGGUGUAUGUCACUCGGCUCGGCCCACUACC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006504
          miRBase ID: mml-miR-668
          Mature miRNA Sequence: UGUCACUCGGCUCGGCCCACUAC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303808_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11867
          Ambion® Pre-miR™ miRNA Precursor : PM11867
          TaqMan™ MicroRNA Assay : 001992
          mirVana® miRNA inhibitor : MH11867
          mirVana® miRNA mimic : MC11867
          MIRNA : TaqMan® OpenArray® Human Advanced MicroRNA Panel
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          FGMIR : TaqManTM Advanced miRNA Human B 96-well Plate 2
          MIRNA : TaqMan® Advanced miRNA Human A and B Cards
          MIRNA : TaqMan® Advanced miRNA Human B 96-well Plates, Standard
          MIRNA : TaqMan® Advanced miRNA Human B Card
          FGMIR : TaqManTM Advanced miRNA Human B Card
          MIRNA : TaqMan® Advanced miRNA Human A and B 96-well Plates, Standard
          FGMIR : QS TaqmanTM OpenArray Human Advanced miRNA Panel

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline