Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12309
          Assay Name: hsa-miR-522-3p
          miRBase Accession Number: MI0003177
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 53751211 - 53751297 [+] on Build GRCh38
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Species: Human, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12309
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000020, mir-515

          Mature miRNA Sequence:

          AAAAUGGUUCCCUUUAGAGUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-522-3p MIMAT0002868
          Gorilla gorilla ggo-miR-522-3p MIMAT0036438
          Pan troglodytes ptr-miR-522 MIMAT0008206
          Pongo pygmaeus ppy-miR-522 MIMAT0015978
          Rhesus monkey mml-miR-518e MIMAT0006382

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-522
          Stem-loop Accession # MI0003177
          Stem-loop Sequence
          UCUCAGGCUGUGUCCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUACGCUUUGAGA
          Chromosome Location Chr. 19 - 53751211 - 53751297 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002868
          miRBase ID: hsa-miR-522-3p
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Chromosome Location: Chr. 19 - 53751211 - 53751297 [+] on Build GRCh38

          Gorilla gorilla

          Stem-loop ID ggo-mir-522
          Stem-loop Accession # MI0031089
          Stem-loop Sequence
          CUGGAGCAAGAAGAUCUCAGGCUGUGUCCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUACGCUUUGAGAAAAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036438
          miRBase ID: ggo-miR-522-3p
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-522
          Stem-loop Accession # MI0008733
          Stem-loop Sequence
          CUCAGGCUGUGUCCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUACGCUUUGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008206
          miRBase ID: ptr-miR-522
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-522
          Stem-loop Accession # MI0015029
          Stem-loop Sequence
          UCUCAGGCUGUGACCCUCUAGAGGGGAGCACUUUCUGUUGUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUAGGCUUUGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015978
          miRBase ID: ppy-miR-522
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-518e
          Stem-loop Accession # MI0007793
          Stem-loop Sequence
          UCUCAGGCUGUGACCCUCUAGAGGGAAGCGAUUUCUGUGAUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUACUGUUUGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006382
          miRBase ID: mml-miR-518e
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs04225384_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12309
          TaqMan™ MicroRNA Assay : 002413
          mirVana® miRNA inhibitor : MH12309
          mirVana® miRNA mimic : MC12309
          TaqMan™ Advanced miRNA Assay : 478993_mir


          miRBase Alias:

          Human: hsa-miR-522 (v17)
          Macaca mulatta: mml-miR-522 (v15)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline