Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12309
          Assay Name: hsa-miR-522-3p
          miRBase Accession Number: MI0003177
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 53751211 - 53751297 [+] on Build GRCh38
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Species: Human, Gorilla gorilla, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC12309

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000020, mir-515

          Mature miRNA Sequence:

          AAAAUGGUUCCCUUUAGAGUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-522-3p MIMAT0002868
          Gorilla gorilla ggo-miR-522-3p MIMAT0036438
          Pan troglodytes ptr-miR-522 MIMAT0008206
          Pongo pygmaeus ppy-miR-522 MIMAT0015978
          Rhesus monkey mml-miR-518e MIMAT0006382

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-522
          Stem-loop Accession # MI0003177
          Stem-loop Sequence
          UCUCAGGCUGUGUCCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUACGCUUUGAGA
          Chromosome Location Chr. 19 - 53751211 - 53751297 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0002868
          miRBase ID: hsa-miR-522-3p
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Chromosome Location: Chr. 19 - 53751211 - 53751297 [+] on Build GRCh38

          Gorilla gorilla

          Stem-loop ID ggo-mir-522
          Stem-loop Accession # MI0031089
          Stem-loop Sequence
          CUGGAGCAAGAAGAUCUCAGGCUGUGUCCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUACGCUUUGAGAAAAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036438
          miRBase ID: ggo-miR-522-3p
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-522
          Stem-loop Accession # MI0008733
          Stem-loop Sequence
          CUCAGGCUGUGUCCCUCUAGAGGGAAGCGCUUUCUGUUGUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUACGCUUUGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008206
          miRBase ID: ptr-miR-522
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-522
          Stem-loop Accession # MI0015029
          Stem-loop Sequence
          UCUCAGGCUGUGACCCUCUAGAGGGGAGCACUUUCUGUUGUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUAGGCUUUGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015978
          miRBase ID: ppy-miR-522
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-518e
          Stem-loop Accession # MI0007793
          Stem-loop Sequence
          UCUCAGGCUGUGACCCUCUAGAGGGAAGCGAUUUCUGUGAUCUGAAAGAAAAGAAAAUGGUUCCCUUUAGAGUGUUACUGUUUGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006382
          miRBase ID: mml-miR-518e
          Mature miRNA Sequence: AAAAUGGUUCCCUUUAGAGUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs04225384_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12309
          Ambion® Pre-miR™ miRNA Precursor : PM12309
          TaqMan™ MicroRNA Assay : 002413
          mirVana® miRNA inhibitor : MH12309
          TaqMan™ Advanced miRNA Assay : 478993_mir


          miRBase Alias:

          Human: hsa-miR-522 (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline