Search
Search

| Assay Name: | hsa-miR-185-5p |
|---|---|
| miRBase Accession Number: | MI0000482 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAGAGAAAGGCAGUUCCUGA |
| Species: | Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Gorilla gorilla, Guinea pig, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000202, mir-185 |
Mature miRNA Sequence: | UGGAGAGAAAGGCAGUUCCUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-185-5p | MIMAT0000455 |
| Mouse | mmu-miR-185-5p | MIMAT0000214 |
| Rat | rno-miR-185-5p | MIMAT0000862 |
| Bovine | bta-miR-185 | MIMAT0009247 |
| Callithrix jacchus | cja-miR-185 | MIMAT0039493 |
| Cricetulus griseus | cgr-miR-185-5p | MIMAT0023811 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-185-5p | MIMAT0047705 |
| Dog | cfa-miR-185 | MIMAT0006660 |
| Gorilla gorilla | ggo-miR-185 | MIMAT0024086 |
| Guinea pig | cpo-miR-185-5p | MIMAT0047054 |
| Pan troglodytes | ptr-miR-185 | MIMAT0008053 |
| Pig | ssc-miR-185 | MIMAT0007759 |
| Pongo pygmaeus | ppy-miR-185 | MIMAT0015781 |
| Pteropus alecto | pal-miR-185-5p | MIMAT0039840 |
| Rabbit | ocu-miR-185-5p | MIMAT0048291 |
| Rhesus monkey | mml-miR-185-5p | MIMAT0006219 |
| Tupaia chinensis | tch-miR-185-5p | MIMAT0036564 |