Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › mmu480964_mir

          Specificity testing was performed using mouse anti-targets.

          Assay Name: mmu-miR-185-5p
          Stem-loop Accession Number: MI0000482
          miRBase Version: v22.1
          Chromosome Location: Chr.22: 20033139 - 20033220 [+] on Build GRCh38
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Gorilla gorilla, Guinea pig, Pan troglodytes, Pig, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey, Tupaia chinensis
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID mmu480964_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000202, mir-185

          Mature miRNA Sequence:

          UGGAGAGAAAGGCAGUUCCUGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-185-5p MIMAT0000455
          Mouse mmu-miR-185-5p MIMAT0000214
          Rat rno-miR-185-5p MIMAT0000862
          Bovine bta-miR-185 MIMAT0009247
          Callithrix jacchus cja-miR-185 MIMAT0039493
          Cricetulus griseus cgr-miR-185-5p MIMAT0023811
          View More Rows
          Dasypus novemcinctus dno-miR-185-5p MIMAT0047705
          Dog cfa-miR-185 MIMAT0006660
          Gorilla gorilla ggo-miR-185 MIMAT0024086
          Guinea pig cpo-miR-185-5p MIMAT0047054
          Pan troglodytes ptr-miR-185 MIMAT0008053
          Pig ssc-miR-185 MIMAT0007759
          Pongo pygmaeus ppy-miR-185 MIMAT0015781
          Pteropus alecto pal-miR-185-5p MIMAT0039840
          Rabbit ocu-miR-185-5p MIMAT0048291
          Rhesus monkey mml-miR-185-5p MIMAT0006219
          Tupaia chinensis tch-miR-185-5p MIMAT0036564

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-185
          Stem-loop Accession # MI0000482
          Stem-loop Sequence
          AGGGGGCGAGGGAUUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCCAGGGGCUGGCUUUCCUCUGGUCCUUCCCUCCCA
          Chromosome Location Chr. 22 - 20033139 - 20033220 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000455
          miRBase ID: hsa-miR-185-5p
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: Chr. 22 - 20033139 - 20033220 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-185
          Stem-loop Accession # MI0000227
          Stem-loop Sequence
          AGGGAUUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCAGGGGCUGGCUUUCCUCUGGUCCUU
          Chromosome Location Chr. 16 - 18327401 - 18327465 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000214
          miRBase ID: mmu-miR-185-5p
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: Chr. 16 - 18327401 - 18327465 [-] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-185
          Stem-loop Accession # MI0000930
          Stem-loop Sequence
          GGGGGUGAGGGAUUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCAGGGGCUGGCUUUCCUCUGGUCCUUCUCUCCCA
          Chromosome Location Chr. 11 - 86812702 - 86812781 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000862
          miRBase ID: rno-miR-185-5p
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: Chr. 11 - 86812702 - 86812781 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-185
          Stem-loop Accession # MI0009758
          Stem-loop Sequence
          GGGGGUGAGGGACUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCCAGGGGCUGGCUUUCCUCCGGCCCCUCCUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009247
          miRBase ID: bta-miR-185
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-185
          Stem-loop Accession # MI0032018
          Stem-loop Sequence
          GGGGGCGAGGGAUUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCCAGGGGCUGGCUUUCCUCUGGUCCUUCCCUCCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039493
          miRBase ID: cja-miR-185
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-185
          Stem-loop Accession # MI0020429
          Stem-loop Sequence
          GUGAGGGAUUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCAGGGGCUGGCUUUCCUCUGGUCCUUCUCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023811
          miRBase ID: cgr-miR-185-5p
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-185
          Stem-loop Accession # MI0039016
          Stem-loop Sequence
          UGGAGAGAAAGGCAGUUCCUGAUGGUGCCUCCCCAGGGGCUGGCUUUCCUCUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047705
          miRBase ID: dno-miR-185-5p
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-185
          Stem-loop Accession # MI0008065
          Stem-loop Sequence
          UGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCCAGGGGCUGGCUUUCCUCUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006660
          miRBase ID: cfa-miR-185
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-185
          Stem-loop Accession # MI0020633
          Stem-loop Sequence
          UCGAGCAGGGGGCGAGGGAUUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCCAGGGGCUGGCUUUCCUCUGGUCCUUCCCUCCCAAUGACCGCGUCUUCGUCGAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024086
          miRBase ID: ggo-miR-185
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-185
          Stem-loop Accession # MI0038673
          Stem-loop Sequence
          UGGAGAGAAAGGCAGUUCCUGAUGGUCCCUUCCCCAGGGGCUGGCUUUCCUCUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047054
          miRBase ID: cpo-miR-185-5p
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-185
          Stem-loop Accession # MI0008561
          Stem-loop Sequence
          GGGGGCGAGGGAUUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCCAGGGGCUGGCUUUCCUCUGGUCCUUCCCUCCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008053
          miRBase ID: ptr-miR-185
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-185
          Stem-loop Accession # MI0008218
          Stem-loop Sequence
          GGUGAGGGAUUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCCAGGGGCUGGCUUUCCUCUGGUCCUUCCCUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007759
          miRBase ID: ssc-miR-185
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-185
          Stem-loop Accession # MI0014849
          Stem-loop Sequence
          AGGGGGCGAGGGAUUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCCAGGGGCUGGCUUUCCUCUGGUCCUUCCCUCCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015781
          miRBase ID: ppy-miR-185
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-185
          Stem-loop Accession # MI0032356
          Stem-loop Sequence
          UGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUGCCCAGGGGCUGGCUUUCCUCUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039840
          miRBase ID: pal-miR-185-5p
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-185
          Stem-loop Accession # MI0039324
          Stem-loop Sequence
          UGGAGAGAAAGGCAGUUCCUGAUGUUCCCCUCCCCAGGGGCUGGCUUUCCUCUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048291
          miRBase ID: ocu-miR-185-5p
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-185
          Stem-loop Accession # MI0007649
          Stem-loop Sequence
          AGGGCGCGAGGGAUUGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCUCAGGGGCUGGCUUUCCUCUGGUCCUUCCCUCCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006219
          miRBase ID: mml-miR-185-5p
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-185
          Stem-loop Accession # MI0031225
          Stem-loop Sequence
          UGGAGAGAAAGGCAGUUCCUGAUGGUCCCCUCCCCAGGGGCUGGCUUUCCUCUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036564
          miRBase ID: tch-miR-185-5p
          Mature miRNA Sequence: UGGAGAGAAAGGCAGUUCCUGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303287_pri
          TaqMan™ Pri-miRNA Assay : Mm03306442_pri
          TaqMan™ Pri-miRNA Assay : Rn03465097_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12486
          Ambion® Pre-miR™ miRNA Precursor : PM12486
          TaqMan™ MicroRNA Assay : 002271
          mirVana® miRNA inhibitor : MH12486
          mirVana® miRNA mimic : MC12486
          TaqMan™ Advanced miRNA Assay : 477939_mir
          TaqMan™ Advanced miRNA Assay : rno480964_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline