Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM12589
          Assay Name: hsa-miR-337-3p
          miRBase Accession Number: MI0000806
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 100874493 - 100874585 [+] on Build GRCh38
          Mature miRNA Sequence: CUCCUAUAUGAUGCCUUUCUUC
          Species: Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM12589
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000195, mir-337

          Mature miRNA Sequence:

          CUCCUAUAUGAUGCCUUUCUUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-337-3p MIMAT0000754
          Pan troglodytes ptr-miR-337 MIMAT0008105
          Pongo pygmaeus ppy-miR-337-3p MIMAT0015839
          Rhesus monkey mml-miR-337-3p MIMAT0006276

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-337
          Stem-loop Accession # MI0000806
          Stem-loop Sequence
          GUAGUCAGUAGUUGGGGGGUGGGAACGGCUUCAUACAGGAGUUGAUGCACAGUUAUCCAGCUCCUAUAUGAUGCCUUUCUUCAUCCCCUUCAA
          Chromosome Location Chr. 14 - 100874493 - 100874585 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000754
          miRBase ID: hsa-miR-337-3p
          Mature miRNA Sequence: CUCCUAUAUGAUGCCUUUCUUC
          Chromosome Location: Chr. 14 - 100874493 - 100874585 [+] on Build GRCh38

          Pan troglodytes

          Stem-loop ID ptr-mir-337
          Stem-loop Accession # MI0008624
          Stem-loop Sequence
          UAGUCAGUAGUUGGGGGGUGGGAACGGCUUCAUACAGGAGUUGAUGCACAGUUAUCCAGCUCCUAUAUGAUGCCUUUCUUCAUCCCCUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008105
          miRBase ID: ptr-miR-337
          Mature miRNA Sequence: CUCCUAUAUGAUGCCUUUCUUC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-337
          Stem-loop Accession # MI0014901
          Stem-loop Sequence
          GUAGUCAGUAGUUGGGGGGUGGGAACGGCUUCAUACAGGAGUUGAUGCACAGUUAUCCAGCUCCUAUAUGAUGCCUUUCUUCAUCCCCUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015839
          miRBase ID: ppy-miR-337-3p
          Mature miRNA Sequence: CUCCUAUAUGAUGCCUUUCUUC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-337
          Stem-loop Accession # MI0007700
          Stem-loop Sequence
          GUAGUCAGUAGUUGGGGGGUGGGAACGGCUUCAUACAGGAGUUGAUGCACAGUUAUCCAGCUCCUAUAUGAUGCCUUUCUUCAUCCCCUUCAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006276
          miRBase ID: mml-miR-337-3p
          Mature miRNA Sequence: CUCCUAUAUGAUGCCUUUCUUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03293725_pri
          Ambion® Pre-miR™ miRNA Precursor : PM12589
          TaqMan™ MicroRNA Assay : 002157
          mirVana® miRNA inhibitor : MH12589
          mirVana® miRNA mimic : MC12589
          TaqMan™ Advanced miRNA Assay : 478035_mir

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0