Search
Search

| Assay Name: | hsa-miR-337-3p |
|---|---|
| miRBase Accession Number: | MI0000806 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCCUAUAUGAUGCCUUUCUUC |
| Species: | Human, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000195, mir-337 |
Mature miRNA Sequence: | CUCCUAUAUGAUGCCUUUCUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-337-3p | MIMAT0000754 |
| Pan troglodytes | ptr-miR-337 | MIMAT0008105 |
| Pongo pygmaeus | ppy-miR-337-3p | MIMAT0015839 |
| Rhesus monkey | mml-miR-337-3p | MIMAT0006276 |