Search
Search

| Assay Name: | hsa-miR-194-3p |
|---|---|
| miRBase Accession Number: | MI0000732 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CCAGUGGGGCUGCUGUUAUCUG |
| Species: | Human, Mouse, Cricetulus griseus, Guinea pig |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000055, mir-194 |
Mature miRNA Sequence: | CCAGUGGGGCUGCUGUUAUCUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-194-3p | MIMAT0004671 |
| Mouse | mmu-miR-194-2-3p | MIMAT0017073 |
| Cricetulus griseus | cgr-miR-194-2 | MIMAT0041134 |
| Guinea pig | cpo-miR-194-3p | MIMAT0047075 |