Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM12608
          Assay Name: hsa-miR-194-3p
          miRBase Accession Number: MI0000732
          miRBase Version: v22.1
          Chromosome Location: Chr.11: 64891355 - 64891439 [-] on Build GRCh38
          Mature miRNA Sequence: CCAGUGGGGCUGCUGUUAUCUG
          Species: Human, Mouse, Cricetulus griseus, Guinea pig
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM12608
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000055, mir-194

          Mature miRNA Sequence:

          CCAGUGGGGCUGCUGUUAUCUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-194-3p MIMAT0004671
          Mouse mmu-miR-194-2-3p MIMAT0017073
          Cricetulus griseus cgr-miR-194-2 MIMAT0041134
          Guinea pig cpo-miR-194-3p MIMAT0047075

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-194-2
          Stem-loop Accession # MI0000732
          Stem-loop Sequence
          UGGUUCCCGCCCCCUGUAACAGCAACUCCAUGUGGAAGUGCCCACUGGUUCCAGUGGGGCUGCUGUUAUCUGGGGCGAGGGCCAG
          Chromosome Location Chr. 11 - 64891355 - 64891439 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004671
          miRBase ID: hsa-miR-194-3p
          Mature miRNA Sequence: CCAGUGGGGCUGCUGUUAUCUG
          Chromosome Location: Chr. 11 - 64891355 - 64891439 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-194-2
          Stem-loop Accession # MI0000733
          Stem-loop Sequence
          GUGGCUCCCACCCUCUGUAACAGCAACUCCAUGUGGAAGUGCCCACUGGUUCCAGUGGGGCUGCUGUUAUCUGGGGUGGCGGCUAG
          Chromosome Location Chr. 19 - 6264643 - 6264728 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017073
          miRBase ID: mmu-miR-194-2-3p
          Mature miRNA Sequence: CCAGUGGGGCUGCUGUUAUCUG
          Chromosome Location: Chr. 19 - 6264643 - 6264728 [+] on Build GRCm38

          Cricetulus griseus

          Stem-loop ID cgr-mir-194-2
          Stem-loop Accession # MI0033062
          Stem-loop Sequence
          UGGCUGCCACCCCCUGUAACAGCAACUCCAUGUGGAAGUGCCCACUAGUUCCAGUGGGGCUGCUGUUAUCUGGGGUGGAGGCUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041134
          miRBase ID: cgr-miR-194-2
          Mature miRNA Sequence: CCAGUGGGGCUGCUGUUAUCUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-194
          Stem-loop Accession # MI0038683
          Stem-loop Sequence
          UGUAACAGCAACUCCAUGUGGAAGUGCCCACUGGUUCCAGUGGGGCUGCUGUUAUCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047075
          miRBase ID: cpo-miR-194-3p
          Mature miRNA Sequence: CCAGUGGGGCUGCUGUUAUCUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302784_pri
          TaqMan™ Pri-miRNA Assay : Mm03306716_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12608
          TaqMan™ MicroRNA Assay : 002379
          mirVana® miRNA inhibitor : MH12608
          mirVana® miRNA mimic : MC12608
          TaqMan™ Advanced miRNA Assay : mmu481600_mir
          TaqMan™ Advanced miRNA Assay : 478743_mir


          miRBase Alias:

          Human: hsa-miR-194* (v17)
          Mouse: mmu-miR-194-2* (v17)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline