Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › AM13025
          Assay Name: hsa-miR-15a-3p
          miRBase Accession Number: MI0000069
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 50049119 - 50049201 [-] on Build GRCh38
          Mature miRNA Sequence: CAGGCCAUAUUGUGCUGCCUCA
          Species: Human, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Monodelphis domestica, Ornithorhynchus anatinus, Rabbit, Taeniopygia guttata
          Product Type: Ambion® Anti-miR™ miRNA Inhibitor
          Assay ID AM13025
          Size
          Availability Made To Order
          Catalog # AM17000
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000006, mir-15

          Mature miRNA Sequence:

          CAGGCCAUAUUGUGCUGCCUCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-15a-3p MIMAT0004488
          Cricetulus griseus cgr-miR-15a-3p MIMAT0023790
          Dasypus novemcinctus dno-miR-15a-3p MIMAT0047502
          Goat chi-miR-15a-3p MIMAT0035991
          Guinea pig cpo-miR-15a-3p MIMAT0046850
          Monodelphis domestica mdo-miR-15a-3p MIMAT0026689
          View More Rows
          Ornithorhynchus anatinus oan-miR-15a-3p MIMAT0007165
          Rabbit ocu-miR-15a-3p MIMAT0048102
          Taeniopygia guttata tgu-miR-15a-3p MIMAT0027000

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-15a
          Stem-loop Accession # MI0000069
          Stem-loop Sequence
          CCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location Chr. 13 - 50049119 - 50049201 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004488
          miRBase ID: hsa-miR-15a-3p
          Mature miRNA Sequence: CAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location: Chr. 13 - 50049119 - 50049201 [-] on Build GRCh38

          Cricetulus griseus

          Stem-loop ID cgr-mir-15a
          Stem-loop Accession # MI0020416
          Stem-loop Sequence
          GGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023790
          miRBase ID: cgr-miR-15a-3p
          Mature miRNA Sequence: CAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-15a
          Stem-loop Accession # MI0038905
          Stem-loop Sequence
          UAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047502
          miRBase ID: dno-miR-15a-3p
          Mature miRNA Sequence: CAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-15a
          Stem-loop Accession # MI0030644
          Stem-loop Sequence
          ACCUUGGAGUAAAGUAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGGAUCUGAUCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035991
          miRBase ID: chi-miR-15a-3p
          Mature miRNA Sequence: CAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-15a
          Stem-loop Accession # MI0038561
          Stem-loop Sequence
          UAGCAGCACAUAAUGGUUUGUGGAUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046850
          miRBase ID: cpo-miR-15a-3p
          Mature miRNA Sequence: CAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-15a
          Stem-loop Accession # MI0005333
          Stem-loop Sequence
          CCUUGGGGUAAAGUAGCAGCACAUAAUGGUUUGUUGGUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026689
          miRBase ID: mdo-miR-15a-3p
          Mature miRNA Sequence: CAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-15a
          Stem-loop Accession # MI0006886
          Stem-loop Sequence
          GAAGACACCUUGGGGUAAAGUAGCAGCACAUAAUGGUUUGUGAGUUUUGAAAUACAGGCCAUAUUGUGCUGCCUCAAAAAUACAAGGAUCUGAUCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007165
          miRBase ID: oan-miR-15a-3p
          Mature miRNA Sequence: CAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-15a
          Stem-loop Accession # MI0039221
          Stem-loop Sequence
          UAGCAGCACAUAAUGGUUUGUGGAGUUUAAAAAGGUGCAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048102
          miRBase ID: ocu-miR-15a-3p
          Mature miRNA Sequence: CAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-15a
          Stem-loop Accession # MI0013772
          Stem-loop Sequence
          UGGCAUAACGUAGCAGCACAUAAUGGUUUGUGGGUUUUGAAAAGGUGCAGGCCAUAUUGUGCUGCCUCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0027000
          miRBase ID: tgu-miR-15a-3p
          Mature miRNA Sequence: CAGGCCAUAUUGUGCUGCCUCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302582_pri
          Ambion® Pre-miR™ miRNA Precursor : PM13025
          TaqMan™ MicroRNA Assay : 002419
          mirVana® miRNA inhibitor : MH13025
          mirVana® miRNA mimic : MC13025
          TaqMan™ Advanced miRNA Assay : 477928_mir


          miRBase Alias:

          Human: hsa-miR-15a* (v17)
          Ochrobactrum anthropi: oan-miR-15a* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline