Search
Search

| Assay Name: | hsa-miR-509-5p |
|---|---|
| miRBase Accession Number: | MI0003196 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UACUGCAGACAGUGGCAAUCA |
| Species: | Human, Horse, Pongo pygmaeus |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | UACUGCAGACAGUGGCAAUCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-509-5p | MIMAT0004779 |
| Horse | eca-miR-509a-5p | MIMAT0013233 |
| Pongo pygmaeus | ppy-miR-509-5p | MIMAT0015933 |