Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC13068
          Assay Name: hsa-miR-509-5p
          miRBase Accession Number: MI0003196
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 147260532 - 147260625 [-] on Build GRCh38
          Mature miRNA Sequence: UACUGCAGACAGUGGCAAUCA
          Species: Human, Horse, Pongo pygmaeus
          Product Type: mirVana® miRNA mimic
          Assay ID MC13068

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000130, mir-506

          Mature miRNA Sequence:

          UACUGCAGACAGUGGCAAUCA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-509-5p MIMAT0004779
          Horse eca-miR-509a-5p MIMAT0013233
          Pongo pygmaeus ppy-miR-509-5p MIMAT0015933

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-509-1
          Stem-loop Accession # MI0003196
          Stem-loop Sequence
          CAUGCUGUGUGUGGUACCCUACUGCAGACAGUGGCAAUCAUGUAUAAUUAAAAAUGAUUGGUACGUCUGUGGGUAGAGUACUGCAUGACACAUG
          Chromosome Location Chr. X - 147260532 - 147260625 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004779
          miRBase ID: hsa-miR-509-5p
          Mature miRNA Sequence: UACUGCAGACAGUGGCAAUCA
          Chromosome Location: Chr. X - 147260532 - 147260625 [-] on Build GRCh38

          Stem-loop ID hsa-mir-509-2
          Stem-loop Accession # MI0005530
          Stem-loop Sequence
          CAUGCUGUGUGUGGUACCCUACUGCAGACAGUGGCAAUCAUGUAUAAUUAAAAAUGAUUGGUACGUCUGUGGGUAGAGUACUGCAUGACAC
          Chromosome Location Chr. X - 147258760 - 147258850 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004779
          miRBase ID: hsa-miR-509-5p
          Mature miRNA Sequence: UACUGCAGACAGUGGCAAUCA
          Chromosome Location: Chr. X - 147260532 - 147260625 [-] on Build GRCh38

          Horse

          Stem-loop ID eca-mir-509a
          Stem-loop Accession # MI0012977
          Stem-loop Sequence
          CAUACUGUGUGUGGUACCCUACUGCAGACAGUGGCAAUCAUGUAUAAUUAAAAAUGAUUGACACCUCUGCAAGUGUAGUAACACAUGACACAUGAGAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013233
          miRBase ID: eca-miR-509a-5p
          Mature miRNA Sequence: UACUGCAGACAGUGGCAAUCA
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-509
          Stem-loop Accession # MI0014982
          Stem-loop Sequence
          CAUGCUGUGUGUGGUACCCUACUGCAGACAGUGGCAAUCAUGUAUAAUUAAAAAUGAUUGGUACGUCUGCAGGUAGAGUACUGCAUGACACAUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015933
          miRBase ID: ppy-miR-509-5p
          Mature miRNA Sequence: UACUGCAGACAGUGGCAAUCA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304192_pri
          TaqMan™ Pri-miRNA Assay : Hs03295863_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM13068
          Ambion® Pre-miR™ miRNA Precursor : PM13068
          TaqMan™ MicroRNA Assay : 002235
          mirVana® miRNA inhibitor : MH13068
          TaqMan™ Advanced miRNA Assay : 478965_mir


          miRBase Alias:

          Enterovirus CA55-1988: eca-miR-509-5p (v20)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline