Search
Search

| Assay Name: | hsa-miR-1290 |
|---|---|
| miRBase Accession Number: | MI0006352 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGAUUUUUGGAUCAGGGA |
| Species: | Human, Pan troglodytes |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000610, mir-1290 |
Mature miRNA Sequence: | UGGAUUUUUGGAUCAGGGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1290 | MIMAT0005880 |
| Pan troglodytes | ptr-miR-1290 | MIMAT0008008 |