Search
Search

| Assay Name: | hsa-miR-449b-3p |
|---|---|
| miRBase Accession Number: | MI0003673 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CAGCCACAACUACCCUGCCACU |
| Species: | Human, Rhesus monkey |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | CAGCCACAACUACCCUGCCACU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-449b-3p | MIMAT0009203 |
| Rhesus monkey | mml-miR-449b-3p | MIMAT0026877 |