Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03304853_pri
          Assay Name:
          hsa-mir-449a
          Stem-loop ID:
          hsa-mir-449a hsa-mir-449b
          Mapped to miRBase Version:
          v22.1
          Chromosome Location:
          Chr.5: 55170532 - 55170622 [-] on Build GRCh38
          Stem-loop Location:
          Chr.5: 55170532 - 55170622 [-] on Build GRCh38 Chr.5: 55170646 - 55170742 [-] on Build GRCh38
          Mature miRNA ID:
          hsa-miR-449a hsa-miR-449b-5p hsa-miR-449b-3p
          Species:
          Human
          Product Type:
          TaqMan™ Pri-miRNA Assay
          Assay ID Hs03304853_pri
          Size
          Availability Made To Order
          Catalog # 4427012
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Amplicon Length:

          108

          Chromosome Location:

          Chr. 5 - 55170245 - 55170245 [-] on 55170245 Build GRCh38

          Species:

          Human

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-449a MIMAT0001541
          Human hsa-miR-449b-5p MIMAT0003327
          Human hsa-miR-449b-3p MIMAT0009203

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-449a
          Stem-loop Accession # MI0001648
          Stem-loop Sequence
          CUGUGUGUGAUGAGCUGGCAGUGUAUUGUUAGCUGGUUGAAUAUGUGAAUGGCAUCGGCUAACAUGCAACUGCUGUCUUAUUGCAUAUACA
          Chromosome Location Chr. 5 - 55170532 - 55170622 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001541
          miRBase ID: hsa-miR-449a
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: Chr. 5 - 55170532 - 55170622 [-] on Build GRCh38

          Gene Family ID MIPF0000133, mir-449
          Distance to Amplicon 244 bases
          Stem-loop ID hsa-mir-449b
          Stem-loop Accession # MI0003673
          Stem-loop Sequence
          UGACCUGAAUCAGGUAGGCAGUGUAUUGUUAGCUGGCUGCUUGGGUCAAGUCAGCAGCCACAACUACCCUGCCACUUGCUUCUGGAUAAAUUCUUCU
          Chromosome Location Chr. 5 - 55170646 - 55170742 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003327
          miRBase ID: hsa-miR-449b-5p
          Mature miRNA Sequence: AGGCAGUGUAUUGUUAGCUGGC
          Chromosome Location: Chr. 5 - 55170646 - 55170742 [-] on Build GRCh38

          miRBase Accession #: MIMAT0009203
          miRBase ID: hsa-miR-449b-3p
          Mature miRNA Sequence: CAGCCACAACUACCCUGCCACU
          Chromosome Location: Chr. 5 - 55170646 - 55170742 [-] on Build GRCh38

          Gene Family ID MIPF0000133, mir-449
          Distance to Amplicon 358 bases

          Back To Top

          More Information




          Related Products

          Ambion® Anti-miR™ miRNA Inhibitor : AM11127
          Ambion® Pre-miR™ miRNA Precursor : PM11127
          TaqMan™ MicroRNA Assay : 001030
          mirVana® miRNA inhibitor : MH11127
          mirVana® miRNA mimic : MC11127
          TaqMan™ Advanced miRNA Assay : rno481132_mir
          TaqMan™ Advanced miRNA Assay : 478561_mir
          TaqMan™ Advanced miRNA Assay : mmu481132_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM11521
          Ambion® Pre-miR™ miRNA Precursor : PM11521
          TaqMan™ MicroRNA Assay : 001608
          mirVana® miRNA inhibitor : MH11521
          mirVana® miRNA mimic : MC11521
          TaqMan™ Advanced miRNA Assay : 479528_mir
          Ambion® Anti-miR™ miRNA Inhibitor : AM15179
          Ambion® Pre-miR™ miRNA Precursor : PM15179
          TaqMan™ MicroRNA Assay : 121215_mat
          mirVana® miRNA inhibitor : MH15179
          mirVana® miRNA mimic : MC15179
          TaqMan™ Advanced miRNA Assay : 478909_mir


          miRBase Alias:

          Human: hsa-mir-449b (v21)

          Back To Top

          Related Products

          • High Capacity RNA-to-cDNA Kit
          • SuperScript® IV VILO™ Master Mix
          • TaqMan® Universal PCR Master Mix
          • TaqMan® Fast Advanced Master Mix
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline