Search
Search

| Assay Name: | hsa-miR-449b-5p |
|---|---|
| miRBase Accession Number: | MI0003673 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGCAGUGUAUUGUUAGCUGGC |
| Species: | Human, Bovine, Goat, Pan troglodytes, Pongo pygmaeus, Rhesus monkey |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | AGGCAGUGUAUUGUUAGCUGGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-449b-5p | MIMAT0003327 |
| Bovine | bta-miR-449b | MIMAT0009321 |
| Goat | chi-miR-449b-5p | MIMAT0036232 |
| Pan troglodytes | ptr-miR-449b | MIMAT0008150 |
| Pongo pygmaeus | ppy-miR-449b | MIMAT0015892 |
| Rhesus monkey | mml-miR-449b-5p | MIMAT0006331 |