Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11521
          Assay Name: hsa-miR-449b-5p
          miRBase Accession Number: MI0003673
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 55170646 - 55170742 [-] on Build GRCh38
          Mature miRNA Sequence: AGGCAGUGUAUUGUUAGCUGGC
          Species: Human, Bovine, Goat, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA mimic
          Assay ID MC11521

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000133, mir-449

          Mature miRNA Sequence:

          AGGCAGUGUAUUGUUAGCUGGC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-449b-5p MIMAT0003327
          Bovine bta-miR-449b MIMAT0009321
          Goat chi-miR-449b-5p MIMAT0036232
          Pan troglodytes ptr-miR-449b MIMAT0008150
          Pongo pygmaeus ppy-miR-449b MIMAT0015892
          Rhesus monkey mml-miR-449b-5p MIMAT0006331

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-449b
          Stem-loop Accession # MI0003673
          Stem-loop Sequence
          UGACCUGAAUCAGGUAGGCAGUGUAUUGUUAGCUGGCUGCUUGGGUCAAGUCAGCAGCCACAACUACCCUGCCACUUGCUUCUGGAUAAAUUCUUCU
          Chromosome Location Chr. 5 - 55170646 - 55170742 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003327
          miRBase ID: hsa-miR-449b-5p
          Mature miRNA Sequence: AGGCAGUGUAUUGUUAGCUGGC
          Chromosome Location: Chr. 5 - 55170646 - 55170742 [-] on Build GRCh38

          Bovine

          Stem-loop ID bta-mir-449b
          Stem-loop Accession # MI0009835
          Stem-loop Sequence
          UGACCUGAAUCAGGUAGGCAGUGUAUUGUUAGCUGGCUGCUUUGGGUCAAUUCAGCAGCCACAACUGCCCUGCCACUUGCUUCUGGAUAAAUUCUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009321
          miRBase ID: bta-miR-449b
          Mature miRNA Sequence: AGGCAGUGUAUUGUUAGCUGGC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-449b
          Stem-loop Accession # MI0030795
          Stem-loop Sequence
          UGACCUGAAUCAGGUAGGCAGUGUAUUGUUAGCUGGCUGCUUUGGGUCAAUUCAGCAGCCACAACUGCCCUGCCACUUGCUUCUGGAUAAAUUCUUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036232
          miRBase ID: chi-miR-449b-5p
          Mature miRNA Sequence: AGGCAGUGUAUUGUUAGCUGGC
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-449b
          Stem-loop Accession # MI0008671
          Stem-loop Sequence
          GACCUGAAUCAGGUAGGCAGUGUAUUGUUAGCUGGCUGCUUGGGUCAAGUCAGCAGCCACAACUACCCUGCCACUUGCUUCUGGAUAAAUUCUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008150
          miRBase ID: ptr-miR-449b
          Mature miRNA Sequence: AGGCAGUGUAUUGUUAGCUGGC
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-449b
          Stem-loop Accession # MI0014948
          Stem-loop Sequence
          UGACCUGAAUCAGGUAGGCAGUGUAUUGUUAGCUGGCUGCUUUGGUCAAGUCAGCAGCCACAACUACCCUGCCACUUGCUUCUGGAUAAAUUCUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015892
          miRBase ID: ppy-miR-449b
          Mature miRNA Sequence: AGGCAGUGUAUUGUUAGCUGGC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-449b
          Stem-loop Accession # MI0007748
          Stem-loop Sequence
          UGACCUGAAUCAGGUAGGCAGUGUAUUGUUAGCUGGCUGCUUGAGUCAAGUCAGCAGCCACAACUACCCUGCCACUUGCUUCUGGAUAAAUUCUUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006331
          miRBase ID: mml-miR-449b-5p
          Mature miRNA Sequence: AGGCAGUGUAUUGUUAGCUGGC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304853_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11521
          Ambion® Pre-miR™ miRNA Precursor : PM11521
          TaqMan™ MicroRNA Assay : 001608
          mirVana® miRNA inhibitor : MH11521
          TaqMan™ Advanced miRNA Assay : 479528_mir


          miRBase Alias:

          Human: hsa-miR-449b (v17)
          Macaca mulatta: mml-miR-449b (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline