Search
Search

| Assay Name: | hsa-miR-449a |
|---|---|
| Stem-loop Accession Number: | MI0001648 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGCAGUGUAUUGUUAGCUGGU |
| Species: | Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Guinea pig, Horse, Monodelphis domestica, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | UGGCAGUGUAUUGUUAGCUGGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-449a | MIMAT0001541 |
| Mouse | mmu-miR-449a-5p | MIMAT0001542 |
| Rat | rno-miR-449a-5p | MIMAT0001543 |
| Bovine | bta-miR-449a | MIMAT0009320 |
| Callithrix jacchus | cja-miR-449a | MIMAT0039495 |
| Cricetulus griseus | cgr-miR-449a | MIMAT0041140 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-449a-5p | MIMAT0047892 |
| Dog | cfa-miR-449a | MIMAT0001544 |
| Guinea pig | cpo-miR-449a-5p | MIMAT0047257 |
| Horse | eca-miR-449a | MIMAT0013102 |
| Monodelphis domestica | mdo-miR-449a-5p | MIMAT0004140 |
| Pongo pygmaeus | ppy-miR-449a | MIMAT0015891 |
| Pteropus alecto | pal-miR-449a-5p | MIMAT0039939 |
| Rabbit | ocu-miR-449a-5p | MIMAT0048453 |
| Rhesus monkey | mml-miR-449a-5p | MIMAT0006330 |