Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › rno481132_mir

          Specificity testing was performed using rat anti-targets.

          Assay Name: rno-miR-449a-5p
          Stem-loop Accession Number: MI0001648
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 55170532 - 55170622 [-] on Build GRCh38
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Species: Human, Mouse, Rat, Bovine, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Guinea pig, Horse, Monodelphis domestica, Pongo pygmaeus, Pteropus alecto, Rabbit, Rhesus monkey
          Product Type: TaqMan™ Advanced miRNA Assay
          Assay ID rno481132_mir
          Size S: 250 rxns
          Availability Inventoried
          Catalog # A25576
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000133, mir-449

          Mature miRNA Sequence:

          UGGCAGUGUAUUGUUAGCUGGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-449a MIMAT0001541
          Mouse mmu-miR-449a-5p MIMAT0001542
          Rat rno-miR-449a-5p MIMAT0001543
          Bovine bta-miR-449a MIMAT0009320
          Callithrix jacchus cja-miR-449a MIMAT0039495
          Cricetulus griseus cgr-miR-449a MIMAT0041140
          View More Rows
          Dasypus novemcinctus dno-miR-449a-5p MIMAT0047892
          Dog cfa-miR-449a MIMAT0001544
          Guinea pig cpo-miR-449a-5p MIMAT0047257
          Horse eca-miR-449a MIMAT0013102
          Monodelphis domestica mdo-miR-449a-5p MIMAT0004140
          Pongo pygmaeus ppy-miR-449a MIMAT0015891
          Pteropus alecto pal-miR-449a-5p MIMAT0039939
          Rabbit ocu-miR-449a-5p MIMAT0048453
          Rhesus monkey mml-miR-449a-5p MIMAT0006330

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-449a
          Stem-loop Accession # MI0001648
          Stem-loop Sequence
          CUGUGUGUGAUGAGCUGGCAGUGUAUUGUUAGCUGGUUGAAUAUGUGAAUGGCAUCGGCUAACAUGCAACUGCUGUCUUAUUGCAUAUACA
          Chromosome Location Chr. 5 - 55170532 - 55170622 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0001541
          miRBase ID: hsa-miR-449a
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: Chr. 5 - 55170532 - 55170622 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-449a
          Stem-loop Accession # MI0001649
          Stem-loop Sequence
          CUGUGUGUGAUGGCUUGGCAGUGUAUUGUUAGCUGGUUGAGUAUGUGAGCGGCACCAGCUAACAUGCGACUGCUCUCCUAUUGCACACACA
          Chromosome Location Chr. 13 - 113037534 - 113037624 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0001542
          miRBase ID: mmu-miR-449a-5p
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: Chr. 13 - 113037534 - 113037624 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-449a
          Stem-loop Accession # MI0001650
          Stem-loop Sequence
          CUGUGUGCGAUGGGUUGGCAGUGUAUUGUUAGCUGGUUGAGUAUGUAAAAGGCACCAGCUAACAUGCAACUGCUCUCCUAUUGCACAUACA
          Chromosome Location Chr. 2 - 44897601 - 44897691 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0001543
          miRBase ID: rno-miR-449a-5p
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: Chr. 2 - 44897601 - 44897691 [+] on Build Rnor_6.0

          Bovine

          Stem-loop ID bta-mir-449a
          Stem-loop Accession # MI0009834
          Stem-loop Sequence
          UGUGUGAUGGGUUGGCAGUGUAUUGUUAGCUGGUUGAAUAUGUGAGUGCCAUCAGCUAACAUGCAACUGCUAUCUUAUUGCAUGUAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009320
          miRBase ID: bta-miR-449a
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-449a
          Stem-loop Accession # MI0032020
          Stem-loop Sequence
          UGUGUGUGAUGAGCUGGCAGUGUAUUGUUAGCUGGUUGAAUAUGUGAAUGGCAUCAGCUAACAUGCAACUGCUGUCUUAUUGCAUAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039495
          miRBase ID: cja-miR-449a
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-449a
          Stem-loop Accession # MI0033069
          Stem-loop Sequence
          CUGUGUGUGACGGGUUGGCAGUGUAUUGUUAGCUGGUUGAAUAUGUGAAAGGCACCAGCUAACAUGCAACUGCUAUCCUGUCGCACAUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041140
          miRBase ID: cgr-miR-449a
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-449a
          Stem-loop Accession # MI0039111
          Stem-loop Sequence
          UGGCAGUGUAUUGUUAGCUGGUUGAAUAUGUGAAUGGCAUCAGCUAACAUUCAGCUGCUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047892
          miRBase ID: dno-miR-449a-5p
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-449a
          Stem-loop Accession # MI0001651
          Stem-loop Sequence
          CCGUGUGUGAUGGGUUGGCAGUGUAUUGUUAGCUGGUUGAAUAUAUGAAUGGCAUCAGCUAACAUGCAACUGCUAUCUUAUUGCAUAUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001544
          miRBase ID: cfa-miR-449a
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-449a
          Stem-loop Accession # MI0038777
          Stem-loop Sequence
          UGGCAGUGUAUUGUUAGCUGGUUGAAUAUAUAUUAUUAGCAGCUAACAUACAACUGCUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047257
          miRBase ID: cpo-miR-449a-5p
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-449a
          Stem-loop Accession # MI0012851
          Stem-loop Sequence
          UGGCAGUGUAUUGUUAGCUGGUUGAAUAUAUGAAUGGCAUCAGCUAACAAGCAACUGCUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013102
          miRBase ID: eca-miR-449a
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-449a
          Stem-loop Accession # MI0005329
          Stem-loop Sequence
          UGUGAUGGGAUGGCAGUGUAUUGUUAGCUGGUUGACUAUCUGAACGUGCCAGCUAACAUGCAACUGCUAUCCCAUUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004140
          miRBase ID: mdo-miR-449a-5p
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-449a
          Stem-loop Accession # MI0014947
          Stem-loop Sequence
          CUGUGUGUGAUGAGCUGGCAGUGUAUUGUUAGCUGGUUGAAUAUGUGAAUGGCAUCGGCUAACGUGCAACUGCUGUCUUAUUGCAUAUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015891
          miRBase ID: ppy-miR-449a
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-449a
          Stem-loop Accession # MI0032410
          Stem-loop Sequence
          UGGCAGUGUAUUGUUAGCUGGUUGAAUAUAUGAAUGGCAUCAGCUAACAUGCAACUGCUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039939
          miRBase ID: pal-miR-449a-5p
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-449a
          Stem-loop Accession # MI0039408
          Stem-loop Sequence
          UGGCAGUGUAUUGUUAGCUGGUUGAAUAUGUGUGUGAUAUCAGCUAACAUACAACUGCUAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048453
          miRBase ID: ocu-miR-449a-5p
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-449a
          Stem-loop Accession # MI0007747
          Stem-loop Sequence
          CUGUGUGUGAUGAGCUGGCAGUGUAUUGUUAGCUGGUUGAAUAUGUGAAUGGCAUCAGCUAACAUGCAACUGCUGUCUUAUUGCAUAUACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006330
          miRBase ID: mml-miR-449a-5p
          Mature miRNA Sequence: UGGCAGUGUAUUGUUAGCUGGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03304853_pri
          TaqMan™ Pri-miRNA Assay : Rn03466118_pri
          TaqMan™ Pri-miRNA Assay : Mm03308638_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11127
          Ambion® Pre-miR™ miRNA Precursor : PM11127
          TaqMan™ MicroRNA Assay : 001030
          mirVana® miRNA inhibitor : MH11127
          mirVana® miRNA mimic : MC11127
          TaqMan™ Advanced miRNA Assay : 478561_mir
          TaqMan™ Advanced miRNA Assay : mmu481132_mir

          Back To Top

          Related Products

          • TaqMan® Advanced miRNA cDNA Synthesis Kit
          • TaqMan® Fast Advanced Master Mix
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline