Search
Search

| Assay Name: | hsa-miR-101-2-5p |
|---|---|
| miRBase Accession Number: | MI0000739 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCGGUUAUCAUGGUACCGAUGC |
| Species: | Human, Rat |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000046, mir-101 |
Mature miRNA Sequence: | UCGGUUAUCAUGGUACCGAUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-101-2-5p | MIMAT0037312 |
| Rat | rno-miR-101b-5p | MIMAT0017045 |